Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19054063-19054324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC6
Entrez Gene ID
93436
Gene Name
armadillo repeat containing 6
Gene Aliases
R30923_1
UniGene
Hs.77876
Gene Chromosome Location
Chr.19: 19033578-19058178 on Build GRCh38
Transcripts
NM_001199196.1 NM_033415.3 XM_005260157.3 XM_017027484.1 XM_017027485.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3100757 c.940G>A p.V314I Chr.19: 19054238-19054238 on Build GRCh38
3530949 c.876G>A p.G292G Chr.19: 19054249-19054249 on Build GRCh38
4663403 c.873_874insG p.L294fs*13 Chr.19: 19054246-19054247 on Build GRCh38
3100759 c.948C>T p.L316L Chr.19: 19054246-19054246 on Build GRCh38
1680815 c.944A>G p.D315G Chr.19: 19054242-19054242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776411936 Chr.19: 19054203-19054203 on Build GRCh38
rs780131030 Chr.19: 19054173-19054173 on Build GRCh38
rs765609632 Chr.19: 19054219-19054219 on Build GRCh38
rs758323734 Chr.19: 19054179-19054179 on Build GRCh38
rs762441082 Chr.19: 19054215-19054215 on Build GRCh38
rs775038322 Chr.19: 19054214-19054214 on Build GRCh38
rs141604457 Chr.19: 19054243-19054243 on Build GRCh38
rs200717495 Chr.19: 19054147-19054147 on Build GRCh38
rs150172819 Chr.19: 19054244-19054244 on Build GRCh38
rs772967762 Chr.19: 19054131-19054131 on Build GRCh38
rs770652073 Chr.19: 19054202-19054202 on Build GRCh38
rs758413432 Chr.19: 19054117-19054117 on Build GRCh38
rs757756285 Chr.19: 19054161-19054161 on Build GRCh38
rs200115218 Chr.19: 19054274-19054274 on Build GRCh38
rs780945141 Chr.19: 19054251-19054251 on Build GRCh38
rs142816550 Chr.19: 19054174-19054174 on Build GRCh38
rs777750459 Chr.19: 19054180-19054180 on Build GRCh38
rs761010517 Chr.19: 19054280-19054280 on Build GRCh38
rs765258191 Chr.19: 19054298-19054298 on Build GRCh38
rs367708399 Chr.19: 19054257-19054257 on Build GRCh38
rs745479381 Chr.19: 19054204-19054204 on Build GRCh38
rs750607969 Chr.19: 19054166-19054166 on Build GRCh38
rs751969684 Chr.19: 19054153-19054153 on Build GRCh38
rs767955278 Chr.19: 19054231-19054231 on Build GRCh38
rs756161107 Chr.19: 19054106-19054106 on Build GRCh38
rs756320781 Chr.19: 19054168-19054168 on Build GRCh38
rs753944717 Chr.19: 19054240-19054240 on Build GRCh38
rs763496559 Chr.19: 19054149-19054149 on Build GRCh38
rs199790725 Chr.19: 19054142-19054142 on Build GRCh38
rs771923785 Chr.19: 19054121-19054121 on Build GRCh38
rs748078025 Chr.19: 19054120-19054120 on Build GRCh38
rs757211966 Chr.19: 19054246-19054246 on Build GRCh38
rs200889331 Chr.19: 19054287-19054287 on Build GRCh38
rs768008307 Chr.19: 19054162-19054162 on Build GRCh38
rs774889530 Chr.19: 19054148-19054148 on Build GRCh38
rs200795233 Chr.19: 19054238-19054238 on Build GRCh38
rs1043927293 Chr.19: 19054247-19054247 on Build GRCh38
rs946868385 Chr.19: 19054246-19054246 on Build GRCh38
rs780003755 Chr.19: 19054112-19054112 on Build GRCh38
rs376530857 Chr.19: 19054132-19054132 on Build GRCh38
rs187591927 Chr.19: 19054146-19054146 on Build GRCh38
rs10414859 Chr.19: 19054273-19054273 on Build GRCh38
rs764551665 Chr.19: 19054150-19054150 on Build GRCh38
rs774941257 Chr.19: 19054220-19054220 on Build GRCh38
rs376186716 Chr.19: 19054227-19054227 on Build GRCh38
rs199499546 Chr.19: 19054198-19054198 on Build GRCh38
rs759756326 Chr.19: 19054292-19054292 on Build GRCh38
rs768216994 Chr.19: 19054271-19054271 on Build GRCh38
rs960339484 Chr.19: 19054276-19054276 on Build GRCh38
rs962544836 Chr.19: 19054181-19054181 on Build GRCh38
rs769383309 Chr.19: 19054206-19054206 on Build GRCh38
rs370794754 Chr.19: 19054241-19054241 on Build GRCh38
rs925739210 Chr.19: 19054084-19054084 on Build GRCh38
rs752825835 Chr.19: 19054304-19054304 on Build GRCh38
rs773595219 Chr.19: 19054279-19054279 on Build GRCh38
rs564011426 Chr.19: 19054306-19054306 on Build GRCh38
rs760781801 Chr.19: 19054233-19054233 on Build GRCh38
rs570950316 Chr.19: 19054105-19054105 on Build GRCh38
rs766606412 Chr.19: 19054282-19054282 on Build GRCh38
rs750773205 Chr.19: 19054237-19054237 on Build GRCh38
rs778287977 Chr.19: 19054161-19054163 on Build GRCh38
rs373212070 Chr.19: 19054136-19054136 on Build GRCh38
rs200535730 Chr.19: 19054265-19054265 on Build GRCh38
rs746788901 Chr.19: 19054186-19054186 on Build GRCh38

View Full Product Details