Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176573860-176574133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDHR2
Entrez Gene ID
54825
Gene Name
cadherin related family member 2
Gene Aliases
PCDH24, PCLKC
UniGene
Hs.4205
Gene Chromosome Location
Chr.5: 176542511-176595768 on Build GRCh38
Transcripts
NM_001171976.1 NM_017675.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4159744 c.449A>T p.D150V Chr.5: 176574126-176574126 on Build GRCh38
3614330 c.435C>T p.S145S Chr.5: 176574112-176574112 on Build GRCh38
6266420 c.448G>T p.D150Y Chr.5: 176574125-176574125 on Build GRCh38
6546958 c.436G>T p.V146L Chr.5: 176574113-176574113 on Build GRCh38
6007184 c.448G>A p.D150N Chr.5: 176574125-176574125 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766304102 Chr.5: 176574101-176574101 on Build GRCh38
rs946263892 Chr.5: 176574017-176574017 on Build GRCh38
rs71601307 Chr.5: 176573909-176573909 on Build GRCh38
rs372367139 Chr.5: 176574085-176574085 on Build GRCh38
rs181246344 Chr.5: 176573907-176573907 on Build GRCh38
rs533948012 Chr.5: 176574024-176574024 on Build GRCh38
rs185777697 Chr.5: 176573915-176573915 on Build GRCh38
rs1030326934 Chr.5: 176573948-176573948 on Build GRCh38
rs552367817 Chr.5: 176573925-176573925 on Build GRCh38
rs754923211 Chr.5: 176574106-176574106 on Build GRCh38
rs1010305754 Chr.5: 176573991-176573991 on Build GRCh38
rs777154668 Chr.5: 176574089-176574089 on Build GRCh38
rs113124612 Chr.5: 176573931-176573931 on Build GRCh38
rs1047535717 Chr.5: 176573916-176573916 on Build GRCh38
rs114719226 Chr.5: 176573921-176573921 on Build GRCh38
rs749454728 Chr.5: 176574040-176574040 on Build GRCh38
rs751436675 Chr.5: 176574103-176574103 on Build GRCh38
rs748008378 Chr.5: 176574111-176574111 on Build GRCh38
rs531835339 Chr.5: 176573952-176573952 on Build GRCh38
rs761268390 Chr.5: 176574073-176574073 on Build GRCh38
rs560289340 Chr.5: 176573920-176573920 on Build GRCh38
rs746006900 Chr.5: 176574059-176574059 on Build GRCh38
rs761329233 Chr.5: 176574091-176574091 on Build GRCh38
rs754265867 Chr.5: 176573891-176573891 on Build GRCh38
rs150841635 Chr.5: 176574092-176574092 on Build GRCh38
rs75866588 Chr.5: 176573985-176573985 on Build GRCh38
rs772442689 Chr.5: 176574066-176574066 on Build GRCh38
rs375699664 Chr.5: 176574057-176574057 on Build GRCh38
rs771208836 Chr.5: 176574042-176574042 on Build GRCh38
rs886593608 Chr.5: 176573919-176573919 on Build GRCh38
rs780918168 Chr.5: 176574107-176574107 on Build GRCh38
rs568482761 Chr.5: 176574001-176574001 on Build GRCh38
rs543816341 Chr.5: 176573887-176573887 on Build GRCh38
rs35424367 Chr.5: 176574099-176574099 on Build GRCh38
rs749881774 Chr.5: 176574096-176574096 on Build GRCh38
rs761988009 Chr.5: 176573897-176573897 on Build GRCh38
rs933502256 Chr.5: 176573966-176573972 on Build GRCh38
rs764491243 Chr.5: 176574088-176574088 on Build GRCh38
rs757392390 Chr.5: 176574113-176574113 on Build GRCh38
rs368192528 Chr.5: 176574067-176574067 on Build GRCh38
rs937125717 Chr.5: 176574053-176574053 on Build GRCh38

View Full Product Details