Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 100985508-100985755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL13A
Entrez Gene ID
392509
Gene Name
ADP ribosylation factor like GTPase 13A
Gene Aliases
ARL13, dJ341D10.2
UniGene
Hs.147237
Gene Chromosome Location
Chr.X: 100969011-100990831 on Build GRCh38
Transcripts
NM_001162491.1 NR_103708.1 NR_103709.1 XM_011530950.2 XM_011530951.2

SNP Detail

SNP ID Chromosome Location
rs757788955 Chr.X: 100985717-100985717 on Build GRCh38
rs111490962 Chr.X: 100985594-100985594 on Build GRCh38
rs758563253 Chr.X: 100985701-100985701 on Build GRCh38
rs776648787 Chr.X: 100985593-100985593 on Build GRCh38
rs768791811 Chr.X: 100985587-100985587 on Build GRCh38
rs760536959 Chr.X: 100985654-100985654 on Build GRCh38
rs758449747 Chr.X: 100985534-100985534 on Build GRCh38
rs752859261 Chr.X: 100985689-100985689 on Build GRCh38
rs987502101 Chr.X: 100985588-100985588 on Build GRCh38
rs199822800 Chr.X: 100985700-100985700 on Build GRCh38
rs767983128 Chr.X: 100985712-100985714 on Build GRCh38
rs369503231 Chr.X: 100985659-100985659 on Build GRCh38
rs747523764 Chr.X: 100985712-100985712 on Build GRCh38
rs1052246293 Chr.X: 100985550-100985550 on Build GRCh38
rs189240515 Chr.X: 100985573-100985573 on Build GRCh38
rs767504263 Chr.X: 100985553-100985553 on Build GRCh38
rs867360172 Chr.X: 100985695-100985695 on Build GRCh38
rs749817740 Chr.X: 100985686-100985687 on Build GRCh38
rs200943867 Chr.X: 100985702-100985702 on Build GRCh38
rs1053962351 Chr.X: 100985616-100985616 on Build GRCh38
rs770001732 Chr.X: 100985627-100985627 on Build GRCh38
rs372025986 Chr.X: 100985664-100985664 on Build GRCh38
rs748429518 Chr.X: 100985615-100985615 on Build GRCh38
rs375350475 Chr.X: 100985688-100985688 on Build GRCh38
rs753623157 Chr.X: 100985667-100985667 on Build GRCh38
rs17257516 Chr.X: 100985586-100985586 on Build GRCh38
rs766195981 Chr.X: 100985663-100985663 on Build GRCh38

View Full Product Details