Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38209082-38209307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG4
Entrez Gene ID
9530
Gene Name
BCL2 associated athanogene 4
Gene Aliases
BAG-4, SODD
UniGene
Hs.194726
Gene Chromosome Location
Chr.8: 38176588-38213301 on Build GRCh38
Transcripts
NM_001204878.1 NM_004874.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3899946 c.758G>A p.G253D Chr.8: 38209137-38209137 on Build GRCh38
7128116 c.796C>A p.P266T Chr.8: 38209175-38209175 on Build GRCh38
4665010 c.800G>A p.G267D Chr.8: 38209179-38209179 on Build GRCh38
5003426 c.760C>T p.R254C Chr.8: 38209139-38209139 on Build GRCh38
4753086 c.782C>T p.A261V Chr.8: 38209161-38209161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772558622 Chr.8: 38209269-38209269 on Build GRCh38
rs771854090 Chr.8: 38209241-38209241 on Build GRCh38
rs756369263 Chr.8: 38209208-38209208 on Build GRCh38
rs768439295 Chr.8: 38209250-38209250 on Build GRCh38
rs140078461 Chr.8: 38209105-38209105 on Build GRCh38
rs531742224 Chr.8: 38209248-38209248 on Build GRCh38
rs773399048 Chr.8: 38209138-38209138 on Build GRCh38
rs761524422 Chr.8: 38209167-38209167 on Build GRCh38
rs749414231 Chr.8: 38209237-38209237 on Build GRCh38
rs768134811 Chr.8: 38209200-38209200 on Build GRCh38
rs766943424 Chr.8: 38209179-38209179 on Build GRCh38
rs749964503 Chr.8: 38209177-38209177 on Build GRCh38
rs769758134 Chr.8: 38209136-38209136 on Build GRCh38
rs760482248 Chr.8: 38209156-38209156 on Build GRCh38
rs772047151 Chr.8: 38209145-38209145 on Build GRCh38
rs781732296 Chr.8: 38209199-38209199 on Build GRCh38
rs752159666 Chr.8: 38209184-38209184 on Build GRCh38
rs550146149 Chr.8: 38209251-38209251 on Build GRCh38
rs759330494 Chr.8: 38209140-38209140 on Build GRCh38
rs748419042 Chr.8: 38209123-38209123 on Build GRCh38
rs755361863 Chr.8: 38209190-38209190 on Build GRCh38
rs376249751 Chr.8: 38209176-38209176 on Build GRCh38
rs561418536 Chr.8: 38209178-38209178 on Build GRCh38
rs775387645 Chr.8: 38209247-38209247 on Build GRCh38
rs201302425 Chr.8: 38209154-38209154 on Build GRCh38
rs202100571 Chr.8: 38209104-38209104 on Build GRCh38
rs775104347 Chr.8: 38209148-38209148 on Build GRCh38
rs143665459 Chr.8: 38209121-38209121 on Build GRCh38
rs777644395 Chr.8: 38209230-38209230 on Build GRCh38
rs146791444 Chr.8: 38209162-38209162 on Build GRCh38

View Full Product Details