Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63752257-63752492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC47
Entrez Gene ID
57003
Gene Name
coiled-coil domain containing 47
Gene Aliases
GK001, MSTP041
UniGene
Hs.202011
Gene Chromosome Location
Chr.17: 63745250-63773728 on Build GRCh38
Transcripts
NM_020198.2 XM_005257527.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
473186 c.1096C>T p.P366S Chr.17: 63752427-63752427 on Build GRCh38
4857867 c.1096C>T p.P366S Chr.17: 63752427-63752427 on Build GRCh38
3820206 c.1149G>T p.M383I Chr.17: 63752374-63752374 on Build GRCh38
6021645 c.1123G>A p.D375N Chr.17: 63752400-63752400 on Build GRCh38
3890157 c.1166C>T p.S389F Chr.17: 63752357-63752357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769204764 Chr.17: 63752379-63752379 on Build GRCh38
rs778458769 Chr.17: 63752439-63752439 on Build GRCh38
rs761325501 Chr.17: 63752370-63752370 on Build GRCh38
rs754719975 Chr.17: 63752301-63752301 on Build GRCh38
rs373538452 Chr.17: 63752294-63752294 on Build GRCh38
rs762011753 Chr.17: 63752286-63752286 on Build GRCh38
rs16947066 Chr.17: 63752432-63752432 on Build GRCh38
rs753941962 Chr.17: 63752284-63752284 on Build GRCh38
rs377436276 Chr.17: 63752311-63752311 on Build GRCh38
rs140293985 Chr.17: 63752346-63752346 on Build GRCh38
rs753300975 Chr.17: 63752433-63752433 on Build GRCh38
rs752674178 Chr.17: 63752333-63752333 on Build GRCh38
rs888256764 Chr.17: 63752357-63752357 on Build GRCh38
rs750604908 Chr.17: 63752404-63752404 on Build GRCh38
rs750123815 Chr.17: 63752378-63752378 on Build GRCh38
rs1039560846 Chr.17: 63752360-63752360 on Build GRCh38
rs972168633 Chr.17: 63752387-63752387 on Build GRCh38
rs370815100 Chr.17: 63752390-63752390 on Build GRCh38
rs910464066 Chr.17: 63752464-63752464 on Build GRCh38
rs757799819 Chr.17: 63752401-63752402 on Build GRCh38
rs746254024 Chr.17: 63752356-63752356 on Build GRCh38
rs151145012 Chr.17: 63752408-63752408 on Build GRCh38
rs756021096 Chr.17: 63752337-63752337 on Build GRCh38
rs777205367 Chr.17: 63752339-63752339 on Build GRCh38
rs745552337 Chr.17: 63752452-63752452 on Build GRCh38
rs144240834 Chr.17: 63752389-63752389 on Build GRCh38
rs763131765 Chr.17: 63752405-63752405 on Build GRCh38
rs748826077 Chr.17: 63752342-63752342 on Build GRCh38
rs560428782 Chr.17: 63752426-63752426 on Build GRCh38
rs747806594 Chr.17: 63752359-63752359 on Build GRCh38
rs775963659 Chr.17: 63752364-63752364 on Build GRCh38
rs765400118 Chr.17: 63752401-63752401 on Build GRCh38
rs778565800 Chr.17: 63752355-63752355 on Build GRCh38
rs756072869 Chr.17: 63752430-63752430 on Build GRCh38
rs765436717 Chr.17: 63752293-63752293 on Build GRCh38

View Full Product Details