Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 38685439-38685700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE8
Entrez Gene ID
144402
Gene Name
copine 8
Gene Aliases
-
UniGene
Hs.40910
Gene Chromosome Location
Chr.12: 38652200-38908593 on Build GRCh38
Transcripts
NM_153634.2 XM_017018851.1 XM_017018852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1202175 c.1215A>C p.K405N Chr.12: 38685546-38685546 on Build GRCh38
1202174 c.1234C>T p.P412S Chr.12: 38685527-38685527 on Build GRCh38
7163389 c.1243T>A p.F415I Chr.12: 38685518-38685518 on Build GRCh38
2206535 c.1151A>G p.N384S Chr.12: 38685610-38685610 on Build GRCh38
6634711 c.1153C>A p.P385T Chr.12: 38685608-38685608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183270935 Chr.12: 38685544-38685544 on Build GRCh38
rs573296580 Chr.12: 38685634-38685635 on Build GRCh38
rs978258344 Chr.12: 38685625-38685625 on Build GRCh38
rs757044788 Chr.12: 38685631-38685631 on Build GRCh38
rs1022340780 Chr.12: 38685565-38685565 on Build GRCh38
rs751325087 Chr.12: 38685486-38685486 on Build GRCh38
rs755854274 Chr.12: 38685468-38685468 on Build GRCh38
rs748836600 Chr.12: 38685635-38685639 on Build GRCh38
rs541704501 Chr.12: 38685627-38685627 on Build GRCh38
rs1050411106 Chr.12: 38685472-38685472 on Build GRCh38
rs777430861 Chr.12: 38685484-38685484 on Build GRCh38
rs769290248 Chr.12: 38685534-38685534 on Build GRCh38
rs757164412 Chr.12: 38685497-38685497 on Build GRCh38
rs766578041 Chr.12: 38685597-38685597 on Build GRCh38
rs755168060 Chr.12: 38685479-38685479 on Build GRCh38
rs199991967 Chr.12: 38685557-38685557 on Build GRCh38
rs781083875 Chr.12: 38685528-38685528 on Build GRCh38
rs944814168 Chr.12: 38685653-38685653 on Build GRCh38
rs767508731 Chr.12: 38685635-38685635 on Build GRCh38
rs755990175 Chr.12: 38685657-38685657 on Build GRCh38
rs745420513 Chr.12: 38685532-38685532 on Build GRCh38
rs889240471 Chr.12: 38685554-38685554 on Build GRCh38
rs774613687 Chr.12: 38685558-38685558 on Build GRCh38
rs765449118 Chr.12: 38685620-38685620 on Build GRCh38
rs568834083 Chr.12: 38685662-38685662 on Build GRCh38
rs374379534 Chr.12: 38685542-38685542 on Build GRCh38
rs760891922 Chr.12: 38685596-38685596 on Build GRCh38
rs201874856 Chr.12: 38685579-38685579 on Build GRCh38
rs759704790 Chr.12: 38685603-38685603 on Build GRCh38
rs748500905 Chr.12: 38685575-38685575 on Build GRCh38
rs776780548 Chr.12: 38685600-38685600 on Build GRCh38
rs773827247 Chr.12: 38685590-38685590 on Build GRCh38
rs967299608 Chr.12: 38685614-38685614 on Build GRCh38
rs371442242 Chr.12: 38685469-38685469 on Build GRCh38
rs750348072 Chr.12: 38685645-38685645 on Build GRCh38
rs989573392 Chr.12: 38685511-38685511 on Build GRCh38

View Full Product Details