Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25103468-25103737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFR3B
Entrez Gene ID
22979
Gene Name
EFR3 homolog B
Gene Aliases
KIAA0953
UniGene
Hs.4892
Gene Chromosome Location
Chr.2: 25042104-25159137 on Build GRCh38
Transcripts
NM_001319099.1 NM_014971.1 XM_011532701.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5624895 c.227C>T p.A76V Chr.2: 25103651-25103651 on Build GRCh38
7203077 c.240G>A p.L80L Chr.2: 25103664-25103664 on Build GRCh38
4435496 c.291G>C p.E97D Chr.2: 25103715-25103715 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs992699785 Chr.2: 25103540-25103540 on Build GRCh38
rs1002552775 Chr.2: 25103675-25103675 on Build GRCh38
rs183968189 Chr.2: 25103501-25103501 on Build GRCh38
rs930926032 Chr.2: 25103576-25103576 on Build GRCh38
rs978365936 Chr.2: 25103604-25103604 on Build GRCh38
rs1009360808 Chr.2: 25103634-25103634 on Build GRCh38
rs1008841126 Chr.2: 25103623-25103623 on Build GRCh38
rs372752080 Chr.2: 25103706-25103706 on Build GRCh38
rs376227733 Chr.2: 25103709-25103709 on Build GRCh38
rs201660815 Chr.2: 25103502-25103502 on Build GRCh38
rs754323247 Chr.2: 25103640-25103640 on Build GRCh38
rs892529462 Chr.2: 25103630-25103630 on Build GRCh38
rs555892323 Chr.2: 25103673-25103673 on Build GRCh38
rs545690984 Chr.2: 25103533-25103533 on Build GRCh38
rs913139468 Chr.2: 25103549-25103549 on Build GRCh38
rs1040452115 Chr.2: 25103624-25103624 on Build GRCh38
rs560491458 Chr.2: 25103547-25103547 on Build GRCh38
rs577057222 Chr.2: 25103610-25103610 on Build GRCh38
rs527647427 Chr.2: 25103603-25103603 on Build GRCh38
rs908261578 Chr.2: 25103518-25103518 on Build GRCh38
rs757740248 Chr.2: 25103652-25103652 on Build GRCh38
rs369391330 Chr.2: 25103616-25103616 on Build GRCh38
rs922398413 Chr.2: 25103703-25103703 on Build GRCh38
rs577893773 Chr.2: 25103510-25103510 on Build GRCh38
rs960131232 Chr.2: 25103535-25103535 on Build GRCh38
rs1036898149 Chr.2: 25103532-25103532 on Build GRCh38
rs942431859 Chr.2: 25103615-25103615 on Build GRCh38
rs538202478 Chr.2: 25103491-25103491 on Build GRCh38
rs889140296 Chr.2: 25103585-25103585 on Build GRCh38
rs1050697052 Chr.2: 25103579-25103579 on Build GRCh38
rs940856706 Chr.2: 25103521-25103521 on Build GRCh38

View Full Product Details