Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 69820831-69821104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf135
Entrez Gene ID
138255
Gene Name
chromosome 9 open reading frame 135
Gene Aliases
-
UniGene
Hs.444459
Gene Chromosome Location
Chr.9: 69820804-69906237 on Build GRCh38
Transcripts
NM_001010940.2 NM_001308084.1 NM_001308085.1 NM_001308086.1 NR_131756.1 NR_131757.1 XM_011518229.2 XM_011518230.2 XM_011518231.2 XM_011518234.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
80949 c.120G>A p.K40K Chr.9: 69820999-69820999 on Build GRCh38
3848700 c.86G>A p.R29H Chr.9: 69820965-69820965 on Build GRCh38
3731050 c.37G>A p.D13N Chr.9: 69820916-69820916 on Build GRCh38
1109743 c.106C>T p.R36C Chr.9: 69820985-69820985 on Build GRCh38
3907746 c.150C>T p.S50S Chr.9: 69821029-69821029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760463078 Chr.9: 69820913-69820913 on Build GRCh38
rs375047820 Chr.9: 69821048-69821048 on Build GRCh38
rs768524639 Chr.9: 69820985-69820985 on Build GRCh38
rs745462491 Chr.9: 69820970-69820970 on Build GRCh38
rs368950631 Chr.9: 69821067-69821067 on Build GRCh38
rs751642816 Chr.9: 69821015-69821015 on Build GRCh38
rs2026796 Chr.9: 69821081-69821081 on Build GRCh38
rs149641101 Chr.9: 69820968-69820968 on Build GRCh38
rs748566007 Chr.9: 69820961-69820961 on Build GRCh38
rs1033327761 Chr.9: 69820889-69820889 on Build GRCh38
rs778216844 Chr.9: 69821058-69821058 on Build GRCh38
rs755057937 Chr.9: 69821020-69821020 on Build GRCh38
rs143163232 Chr.9: 69820910-69820910 on Build GRCh38
rs768170659 Chr.9: 69821080-69821080 on Build GRCh38
rs753690197 Chr.9: 69820924-69820924 on Build GRCh38
rs746539745 Chr.9: 69821078-69821078 on Build GRCh38
rs755499542 Chr.9: 69820959-69820959 on Build GRCh38
rs774119117 Chr.9: 69820882-69820882 on Build GRCh38
rs747552571 Chr.9: 69820954-69820954 on Build GRCh38
rs753210256 Chr.9: 69821033-69821033 on Build GRCh38
rs747761453 Chr.9: 69821079-69821080 on Build GRCh38
rs750600894 Chr.9: 69820928-69820928 on Build GRCh38
rs781711958 Chr.9: 69820960-69820960 on Build GRCh38
rs778571754 Chr.9: 69820950-69820951 on Build GRCh38
rs779735236 Chr.9: 69821068-69821068 on Build GRCh38
rs138120161 Chr.9: 69820867-69820867 on Build GRCh38
rs766889158 Chr.9: 69820902-69820902 on Build GRCh38
rs1046347114 Chr.9: 69821034-69821034 on Build GRCh38
rs764088063 Chr.9: 69820916-69820916 on Build GRCh38
rs781582859 Chr.9: 69821028-69821028 on Build GRCh38
rs760218675 Chr.9: 69820979-69820979 on Build GRCh38
rs61146180 Chr.9: 69820876-69820876 on Build GRCh38
rs779936251 Chr.9: 69820937-69820937 on Build GRCh38
rs370333172 Chr.9: 69820915-69820915 on Build GRCh38
rs867663881 Chr.9: 69820942-69820942 on Build GRCh38
rs755324521 Chr.9: 69820884-69820885 on Build GRCh38
rs780639954 Chr.9: 69820856-69820856 on Build GRCh38
rs373790286 Chr.9: 69821004-69821004 on Build GRCh38
rs758492391 Chr.9: 69820929-69820929 on Build GRCh38
rs759150916 Chr.9: 69820883-69820883 on Build GRCh38
rs757022096 Chr.9: 69820925-69820925 on Build GRCh38
rs150197009 Chr.9: 69820972-69820972 on Build GRCh38
rs868544514 Chr.9: 69820880-69820880 on Build GRCh38
rs769075775 Chr.9: 69820875-69820875 on Build GRCh38
rs770759896 Chr.9: 69820877-69820877 on Build GRCh38
rs761626492 Chr.9: 69820990-69820990 on Build GRCh38
rs766616336 Chr.9: 69821012-69821012 on Build GRCh38
rs750655220 Chr.9: 69820997-69820997 on Build GRCh38
rs138857531 Chr.9: 69820978-69820978 on Build GRCh38
rs148273758 Chr.9: 69820926-69820926 on Build GRCh38

View Full Product Details