Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30287765-30288030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLMH
Entrez Gene ID
642
Gene Name
bleomycin hydrolase
Gene Aliases
BH, BMH
UniGene
Hs.371914
Gene Chromosome Location
Chr.17: 30248195-30292166 on Build GRCh38
Transcripts
NM_000386.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3988901 c.350C>T p.A117V Chr.17: 30287919-30287919 on Build GRCh38
1381951 c.329G>A p.R110H Chr.17: 30287940-30287940 on Build GRCh38
4852382 c.339C>T p.F113F Chr.17: 30287930-30287930 on Build GRCh38
401693 c.355G>C p.V119L Chr.17: 30287914-30287914 on Build GRCh38
4493577 c.417C>T p.N139N Chr.17: 30287852-30287852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777647901 Chr.17: 30287902-30287903 on Build GRCh38
rs754251135 Chr.17: 30287802-30287802 on Build GRCh38
rs978088535 Chr.17: 30287796-30287796 on Build GRCh38
rs766858828 Chr.17: 30287800-30287800 on Build GRCh38
rs765650218 Chr.17: 30287941-30287941 on Build GRCh38
rs773407348 Chr.17: 30287906-30287906 on Build GRCh38
rs749998835 Chr.17: 30287963-30287963 on Build GRCh38
rs978305089 Chr.17: 30287797-30287797 on Build GRCh38
rs552944635 Chr.17: 30287841-30287841 on Build GRCh38
rs201644080 Chr.17: 30287982-30287982 on Build GRCh38
rs751251058 Chr.17: 30287809-30287809 on Build GRCh38
rs759989325 Chr.17: 30287934-30287934 on Build GRCh38
rs916736219 Chr.17: 30288000-30288000 on Build GRCh38
rs200618986 Chr.17: 30287814-30287814 on Build GRCh38
rs367865064 Chr.17: 30287833-30287833 on Build GRCh38
rs779669377 Chr.17: 30287985-30287985 on Build GRCh38
rs371722422 Chr.17: 30287923-30287923 on Build GRCh38
rs754375855 Chr.17: 30287924-30287924 on Build GRCh38
rs772118214 Chr.17: 30287887-30287887 on Build GRCh38
rs760721245 Chr.17: 30287922-30287922 on Build GRCh38
rs780698812 Chr.17: 30287962-30287962 on Build GRCh38
rs752941039 Chr.17: 30287955-30287955 on Build GRCh38
rs749259171 Chr.17: 30287994-30287994 on Build GRCh38
rs749133129 Chr.17: 30287868-30287868 on Build GRCh38
rs768600658 Chr.17: 30287871-30287871 on Build GRCh38
rs140020352 Chr.17: 30287831-30287831 on Build GRCh38
rs189020219 Chr.17: 30287884-30287884 on Build GRCh38
rs200146621 Chr.17: 30287973-30287973 on Build GRCh38
rs773966875 Chr.17: 30287878-30287878 on Build GRCh38
rs769906453 Chr.17: 30287839-30287839 on Build GRCh38
rs991388924 Chr.17: 30287939-30287939 on Build GRCh38
rs779108864 Chr.17: 30287804-30287804 on Build GRCh38

View Full Product Details