Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 41361780-41362031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMILIN3
Entrez Gene ID
90187
Gene Name
elastin microfibril interfacer 3
Gene Aliases
C20orf130, EMILIN5, dJ620E11.4
UniGene
Hs.726525
Gene Chromosome Location
Chr.20: 41359966-41366858 on Build GRCh38
Transcripts
NM_052846.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
362126 c.1724G>T p.G575V Chr.20: 41361845-41361845 on Build GRCh38
5013599 c.1783A>G p.S595G Chr.20: 41361786-41361786 on Build GRCh38
3291565 c.1663G>A p.A555T Chr.20: 41361906-41361906 on Build GRCh38
4795934 c.1563C>T p.C521C Chr.20: 41362006-41362006 on Build GRCh38
6467682 c.1629G>A p.R543R Chr.20: 41361940-41361940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2235592 Chr.20: 41361974-41361974 on Build GRCh38
rs550076465 Chr.20: 41361898-41361898 on Build GRCh38
rs763882899 Chr.20: 41361961-41361961 on Build GRCh38
rs745493571 Chr.20: 41361943-41361943 on Build GRCh38
rs747443459 Chr.20: 41361842-41361842 on Build GRCh38
rs139770210 Chr.20: 41361942-41361942 on Build GRCh38
rs963809181 Chr.20: 41362010-41362010 on Build GRCh38
rs750293313 Chr.20: 41361980-41361980 on Build GRCh38
rs776818748 Chr.20: 41361852-41361852 on Build GRCh38
rs113190238 Chr.20: 41361841-41361841 on Build GRCh38
rs777404496 Chr.20: 41361819-41361819 on Build GRCh38
rs950857210 Chr.20: 41361998-41361998 on Build GRCh38
rs375896742 Chr.20: 41361860-41361860 on Build GRCh38
rs142261892 Chr.20: 41361870-41361870 on Build GRCh38
rs747136348 Chr.20: 41361925-41361925 on Build GRCh38
rs139544093 Chr.20: 41361906-41361906 on Build GRCh38
rs774820231 Chr.20: 41361955-41361955 on Build GRCh38
rs764125854 Chr.20: 41361884-41361884 on Build GRCh38
rs151073469 Chr.20: 41361940-41361940 on Build GRCh38
rs762168753 Chr.20: 41361854-41361854 on Build GRCh38
rs144312180 Chr.20: 41361956-41361956 on Build GRCh38
rs779977207 Chr.20: 41361923-41361923 on Build GRCh38
rs189552689 Chr.20: 41361913-41361913 on Build GRCh38
rs768976200 Chr.20: 41361931-41361931 on Build GRCh38
rs781195826 Chr.20: 41362001-41362001 on Build GRCh38
rs752297727 Chr.20: 41361874-41361874 on Build GRCh38
rs949345254 Chr.20: 41361851-41361851 on Build GRCh38
rs149625251 Chr.20: 41361808-41361808 on Build GRCh38
rs199822264 Chr.20: 41361992-41361992 on Build GRCh38
rs8182638 Chr.20: 41361979-41361979 on Build GRCh38
rs200581119 Chr.20: 41361970-41361970 on Build GRCh38
rs144193156 Chr.20: 41361936-41361936 on Build GRCh38
rs757135035 Chr.20: 41361900-41361900 on Build GRCh38
rs149785434 Chr.20: 41361969-41361969 on Build GRCh38
rs758646920 Chr.20: 41361985-41361985 on Build GRCh38
rs770942609 Chr.20: 41361823-41361823 on Build GRCh38
rs990336019 Chr.20: 41361814-41361814 on Build GRCh38
rs745889445 Chr.20: 41361829-41361829 on Build GRCh38
rs772266071 Chr.20: 41361833-41361833 on Build GRCh38
rs766626679 Chr.20: 41361991-41361991 on Build GRCh38
rs771775276 Chr.20: 41361954-41361954 on Build GRCh38
rs770139052 Chr.20: 41361941-41361941 on Build GRCh38
rs61739308 Chr.20: 41362009-41362009 on Build GRCh38
rs866307238 Chr.20: 41361924-41361924 on Build GRCh38
rs114022947 Chr.20: 41361805-41361805 on Build GRCh38
rs995441205 Chr.20: 41361934-41361934 on Build GRCh38
rs745810312 Chr.20: 41361871-41361871 on Build GRCh38
rs368864313 Chr.20: 41361907-41361907 on Build GRCh38
rs1018372793 Chr.20: 41361948-41361948 on Build GRCh38
rs753687591 Chr.20: 41361822-41361822 on Build GRCh38
rs146385182 Chr.20: 41361889-41361889 on Build GRCh38
rs999349926 Chr.20: 41361933-41361933 on Build GRCh38
rs755071876 Chr.20: 41362000-41362000 on Build GRCh38
rs781728666 Chr.20: 41361937-41361937 on Build GRCh38
rs773979845 Chr.20: 41361861-41361861 on Build GRCh38

View Full Product Details