Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1641284-1641527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM53A
Entrez Gene ID
152877
Gene Name
family with sequence similarity 53 member A
Gene Aliases
DNTNP
UniGene
Hs.143314
Gene Chromosome Location
Chr.4: 1574055-1684313 on Build GRCh38
Transcripts
NM_001013622.3 NM_001174070.2 NM_001297435.1 XM_005247948.3 XM_011513402.2 XM_011513403.1 XM_011513404.2 XM_011513405.1 XM_017007802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6999816 c.1112C>T p.P371L Chr.4: 1641378-1641378 on Build GRCh38
3601594 c.1146C>T p.V382V Chr.4: 1641344-1641344 on Build GRCh38
4653786 c.981C>T p.S327S Chr.4: 1641509-1641509 on Build GRCh38
6859908 c.1029C>T p.L343L Chr.4: 1641461-1641461 on Build GRCh38
4683067 c.1074G>A p.S358S Chr.4: 1641416-1641416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370542055 Chr.4: 1641476-1641476 on Build GRCh38
rs759997946 Chr.4: 1641327-1641327 on Build GRCh38
rs750169567 Chr.4: 1641341-1641341 on Build GRCh38
rs370176108 Chr.4: 1641329-1641329 on Build GRCh38
rs532835490 Chr.4: 1641333-1641333 on Build GRCh38
rs1035074252 Chr.4: 1641354-1641354 on Build GRCh38
rs200406395 Chr.4: 1641311-1641311 on Build GRCh38
rs751213329 Chr.4: 1641416-1641416 on Build GRCh38
rs563061445 Chr.4: 1641359-1641359 on Build GRCh38
rs115079906 Chr.4: 1641376-1641376 on Build GRCh38
rs144405478 Chr.4: 1641477-1641477 on Build GRCh38
rs1049019214 Chr.4: 1641368-1641368 on Build GRCh38
rs755718686 Chr.4: 1641438-1641438 on Build GRCh38
rs747415692 Chr.4: 1641475-1641475 on Build GRCh38
rs545751174 Chr.4: 1641322-1641322 on Build GRCh38
rs754643484 Chr.4: 1641417-1641417 on Build GRCh38
rs1022202820 Chr.4: 1641362-1641362 on Build GRCh38
rs779225933 Chr.4: 1641451-1641451 on Build GRCh38
rs775759980 Chr.4: 1641330-1641330 on Build GRCh38
rs746513666 Chr.4: 1641312-1641312 on Build GRCh38
rs200678397 Chr.4: 1641479-1641479 on Build GRCh38
rs758641802 Chr.4: 1641466-1641466 on Build GRCh38
rs566928749 Chr.4: 1641397-1641397 on Build GRCh38
rs749831219 Chr.4: 1641321-1641321 on Build GRCh38
rs772970772 Chr.4: 1641505-1641505 on Build GRCh38
rs778363690 Chr.4: 1641319-1641319 on Build GRCh38
rs748109680 Chr.4: 1641486-1641486 on Build GRCh38
rs147463608 Chr.4: 1641379-1641379 on Build GRCh38
rs762009470 Chr.4: 1641386-1641386 on Build GRCh38
rs956408187 Chr.4: 1641409-1641409 on Build GRCh38
rs565614356 Chr.4: 1641331-1641331 on Build GRCh38
rs768684936 Chr.4: 1641382-1641382 on Build GRCh38
rs751266708 Chr.4: 1641347-1641347 on Build GRCh38
rs781037566 Chr.4: 1641421-1641421 on Build GRCh38
rs199799761 Chr.4: 1641372-1641372 on Build GRCh38
rs780429865 Chr.4: 1641467-1641467 on Build GRCh38
rs779534648 Chr.4: 1641375-1641375 on Build GRCh38
rs147613570 Chr.4: 1641436-1641436 on Build GRCh38
rs145316076 Chr.4: 1641433-1641433 on Build GRCh38
rs769097428 Chr.4: 1641434-1641434 on Build GRCh38
rs761712884 Chr.4: 1641450-1641450 on Build GRCh38
rs764172167 Chr.4: 1641332-1641332 on Build GRCh38
rs150651838 Chr.4: 1641307-1641307 on Build GRCh38
rs373049780 Chr.4: 1641369-1641369 on Build GRCh38
rs745494459 Chr.4: 1641305-1641305 on Build GRCh38
rs1022091487 Chr.4: 1641443-1641443 on Build GRCh38
rs775633001 Chr.4: 1641378-1641378 on Build GRCh38
rs779785935 Chr.4: 1641346-1641346 on Build GRCh38
rs769929489 Chr.4: 1641487-1641487 on Build GRCh38
rs376155315 Chr.4: 1641383-1641383 on Build GRCh38
rs199624823 Chr.4: 1641358-1641358 on Build GRCh38
rs139732204 Chr.4: 1641388-1641388 on Build GRCh38
rs998006102 Chr.4: 1641395-1641395 on Build GRCh38
rs762974379 Chr.4: 1641507-1641507 on Build GRCh38
rs926863953 Chr.4: 1641494-1641494 on Build GRCh38
rs765267471 Chr.4: 1641337-1641337 on Build GRCh38
rs765066869 Chr.4: 1641502-1641502 on Build GRCh38
rs766456718 Chr.4: 1641508-1641508 on Build GRCh38
rs374029502 Chr.4: 1641498-1641498 on Build GRCh38
rs776409657 Chr.4: 1641336-1641337 on Build GRCh38
rs761689023 Chr.4: 1641336-1641336 on Build GRCh38
rs202151764 Chr.4: 1641396-1641396 on Build GRCh38
rs267600065 Chr.4: 1641392-1641392 on Build GRCh38
rs527699371 Chr.4: 1641463-1641463 on Build GRCh38

View Full Product Details