Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111229745-111229992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHI3L2
Entrez Gene ID
1117
Gene Name
chitinase 3 like 2
Gene Aliases
CHIL2, YKL-39, YKL39
UniGene
Hs.514840
Gene Chromosome Location
Chr.1: 111227659-111243440 on Build GRCh38
Transcripts
NM_001025197.1 NM_001025199.1 NM_004000.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5887543 c.68G>A p.G23E Chr.1: 111229879-111229879 on Build GRCh38
6120582 c.43G>A p.V15I Chr.1: 111229854-111229854 on Build GRCh38
5477435 c.42T>G p.G14G Chr.1: 111229853-111229853 on Build GRCh38
893774 c.65A>G p.Q22R Chr.1: 111229876-111229876 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs888961597 Chr.1: 111229854-111229854 on Build GRCh38
rs781315999 Chr.1: 111229862-111229862 on Build GRCh38
rs767003501 Chr.1: 111229892-111229892 on Build GRCh38
rs560709708 Chr.1: 111229781-111229781 on Build GRCh38
rs749883055 Chr.1: 111229894-111229894 on Build GRCh38
rs529738396 Chr.1: 111229783-111229783 on Build GRCh38
rs770961252 Chr.1: 111229859-111229859 on Build GRCh38
rs368680461 Chr.1: 111229946-111229946 on Build GRCh38
rs778084537 Chr.1: 111229931-111229931 on Build GRCh38
rs994196548 Chr.1: 111229943-111229943 on Build GRCh38
rs778816880 Chr.1: 111229821-111229836 on Build GRCh38
rs765201766 Chr.1: 111229942-111229942 on Build GRCh38
rs1039642291 Chr.1: 111229958-111229958 on Build GRCh38
rs1025626263 Chr.1: 111229944-111229944 on Build GRCh38
rs762786574 Chr.1: 111229884-111229884 on Build GRCh38
rs373136804 Chr.1: 111229838-111229838 on Build GRCh38
rs941817165 Chr.1: 111229868-111229868 on Build GRCh38
rs774479360 Chr.1: 111229886-111229886 on Build GRCh38
rs1050739566 Chr.1: 111229829-111229829 on Build GRCh38
rs761797002 Chr.1: 111229890-111229890 on Build GRCh38
rs752060289 Chr.1: 111229933-111229933 on Build GRCh38
rs901999567 Chr.1: 111229966-111229966 on Build GRCh38
rs770072900 Chr.1: 111229879-111229879 on Build GRCh38
rs753095509 Chr.1: 111229915-111229915 on Build GRCh38
rs746141602 Chr.1: 111229869-111229869 on Build GRCh38
rs758806164 Chr.1: 111229929-111229929 on Build GRCh38
rs926474363 Chr.1: 111229790-111229790 on Build GRCh38
rs1043962010 Chr.1: 111229935-111229935 on Build GRCh38
rs768744068 Chr.1: 111229885-111229885 on Build GRCh38
rs369491317 Chr.1: 111229905-111229905 on Build GRCh38
rs879136285 Chr.1: 111229947-111229947 on Build GRCh38
rs992184301 Chr.1: 111229880-111229880 on Build GRCh38
rs778217341 Chr.1: 111229810-111229810 on Build GRCh38

View Full Product Details