Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171453153-171453396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF17
Entrez Gene ID
80067
Gene Name
DDB1 and CUL4 associated factor 17
Gene Aliases
C2orf37
UniGene
Hs.659439
Gene Chromosome Location
Chr.2: 171434166-171491029 on Build GRCh38
Transcripts
NM_001164821.1 NM_025000.3 NR_028482.1 XM_006712767.1 XM_006712768.1 XM_011511881.1 XM_011511882.1 XM_017004995.1 XM_017004996.1 XM_017004997.1 XM_017004998.1 XM_017004999.1 XM_017005000.1 XM_017005001.1 XM_017005002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4818477 c.613G>C p.E205Q Chr.2: 171453199-171453199 on Build GRCh38
1009537 c.619A>G p.N207D Chr.2: 171453205-171453205 on Build GRCh38
7411655 c.606G>A p.G202G Chr.2: 171453192-171453192 on Build GRCh38
5860592 c.591delT p.S199fs*3 Chr.2: 171453177-171453177 on Build GRCh38
3391117 c.596C>T p.S199L Chr.2: 171453182-171453182 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778114154 Chr.2: 171453176-171453176 on Build GRCh38
rs909891679 Chr.2: 171453367-171453367 on Build GRCh38
rs1006857807 Chr.2: 171453303-171453303 on Build GRCh38
rs558760770 Chr.2: 171453291-171453291 on Build GRCh38
rs536507350 Chr.2: 171453208-171453208 on Build GRCh38
rs774670596 Chr.2: 171453263-171453263 on Build GRCh38
rs949562441 Chr.2: 171453226-171453226 on Build GRCh38
rs573074710 Chr.2: 171453282-171453282 on Build GRCh38
rs999240326 Chr.2: 171453274-171453278 on Build GRCh38
rs757257068 Chr.2: 171453194-171453194 on Build GRCh38
rs189836615 Chr.2: 171453283-171453283 on Build GRCh38
rs902160806 Chr.2: 171453187-171453187 on Build GRCh38
rs374256774 Chr.2: 171453230-171453230 on Build GRCh38
rs985084045 Chr.2: 171453354-171453354 on Build GRCh38
rs753906990 Chr.2: 171453177-171453177 on Build GRCh38
rs199780746 Chr.2: 171453259-171453259 on Build GRCh38
rs924087179 Chr.2: 171453270-171453270 on Build GRCh38
rs780253154 Chr.2: 171453241-171453241 on Build GRCh38
rs1018688499 Chr.2: 171453276-171453279 on Build GRCh38
rs772669685 Chr.2: 171453232-171453232 on Build GRCh38
rs747460553 Chr.2: 171453255-171453255 on Build GRCh38
rs879401359 Chr.2: 171453315-171453315 on Build GRCh38
rs778975572 Chr.2: 171453197-171453197 on Build GRCh38

View Full Product Details