Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31531011-31531284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1G2-DDX39B
Entrez Gene ID
100532737
Gene Name
ATP6V1G2-DDX39B readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.254042
Gene Chromosome Location
Chr.6: 31530219-31546848 on Build GRCh38
Transcripts
NR_037853.1 NM_004640.6 NM_080598.5 NR_037852.1

Gene Symbol
DDX39B
Entrez Gene ID
7919
Gene Name
DEAD-box helicase 39B
Gene Aliases
BAT1, D6S81E, UAP56
UniGene
Hs.254042
Gene Chromosome Location
Chr.6: 31530219-31542475 on Build GRCh38
Transcripts
NR_037853.1 NM_004640.6 NM_080598.5 NR_037852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4859163 c.1066C>A p.L356I Chr.6: 31531154-31531154 on Build GRCh38
5563617 c.1009C>G p.Q337E Chr.6: 31531166-31531166 on Build GRCh38
5888897 c.1076C>A p.T359N Chr.6: 31531144-31531144 on Build GRCh38
5870151 c.1003G>A p.D335N Chr.6: 31531172-31531172 on Build GRCh38
3830102 c.1119T>C p.I373I Chr.6: 31531101-31531101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746179453 Chr.6: 31531065-31531065 on Build GRCh38
rs770168629 Chr.6: 31531067-31531067 on Build GRCh38
rs746343032 Chr.6: 31531229-31531229 on Build GRCh38
rs138849178 Chr.6: 31531137-31531137 on Build GRCh38
rs754920476 Chr.6: 31531144-31531144 on Build GRCh38
rs746480137 Chr.6: 31531190-31531190 on Build GRCh38
rs201968778 Chr.6: 31531086-31531086 on Build GRCh38
rs372300280 Chr.6: 31531167-31531167 on Build GRCh38
rs780726042 Chr.6: 31531224-31531224 on Build GRCh38
rs756778715 Chr.6: 31531210-31531210 on Build GRCh38
rs983399281 Chr.6: 31531251-31531252 on Build GRCh38
rs746582265 Chr.6: 31531038-31531038 on Build GRCh38
rs758059764 Chr.6: 31531170-31531170 on Build GRCh38
rs749625838 Chr.6: 31531246-31531246 on Build GRCh38
rs755218640 Chr.6: 31531116-31531116 on Build GRCh38
rs1001010377 Chr.6: 31531062-31531062 on Build GRCh38
rs775758603 Chr.6: 31531068-31531068 on Build GRCh38
rs750186987 Chr.6: 31531134-31531137 on Build GRCh38
rs1006873302 Chr.6: 31531061-31531061 on Build GRCh38
rs765054898 Chr.6: 31531161-31531161 on Build GRCh38
rs1018803787 Chr.6: 31531110-31531110 on Build GRCh38
rs370219054 Chr.6: 31531111-31531111 on Build GRCh38
rs775909387 Chr.6: 31531243-31531243 on Build GRCh38
rs777307555 Chr.6: 31531189-31531189 on Build GRCh38
rs771358976 Chr.6: 31531039-31531039 on Build GRCh38
rs1017404759 Chr.6: 31531089-31531089 on Build GRCh38
rs777151694 Chr.6: 31531042-31531042 on Build GRCh38
rs770270204 Chr.6: 31531235-31531235 on Build GRCh38
rs761990213 Chr.6: 31531125-31531125 on Build GRCh38

View Full Product Details