Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35459579-35459838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCE
Entrez Gene ID
2178
Gene Name
Fanconi anemia complementation group E
Gene Aliases
FACE, FAE
UniGene
Hs.302003
Gene Chromosome Location
Chr.6: 35452339-35467106 on Build GRCh38
Transcripts
NM_021922.2 XM_005248885.2 XM_005248886.2 XM_005248887.2 XM_005248888.3 XM_011514343.2

SNP Detail

SNP ID Chromosome Location
rs758380315 Chr.6: 35459722-35459722 on Build GRCh38
rs767772525 Chr.6: 35459614-35459614 on Build GRCh38
rs774433312 Chr.6: 35459650-35459650 on Build GRCh38
rs45571538 Chr.6: 35459647-35459647 on Build GRCh38
rs574832072 Chr.6: 35459649-35459649 on Build GRCh38
rs530543374 Chr.6: 35459809-35459809 on Build GRCh38
rs914519306 Chr.6: 35459791-35459791 on Build GRCh38
rs886061331 Chr.6: 35459734-35459734 on Build GRCh38
rs747692546 Chr.6: 35459707-35459707 on Build GRCh38
rs1013143533 Chr.6: 35459700-35459700 on Build GRCh38
rs777907301 Chr.6: 35459748-35459748 on Build GRCh38
rs761282617 Chr.6: 35459651-35459651 on Build GRCh38
rs956841290 Chr.6: 35459613-35459613 on Build GRCh38
rs200479051 Chr.6: 35459770-35459770 on Build GRCh38
rs559435652 Chr.6: 35459690-35459690 on Build GRCh38
rs927227725 Chr.6: 35459680-35459680 on Build GRCh38
rs776298967 Chr.6: 35459760-35459760 on Build GRCh38
rs761927217 Chr.6: 35459798-35459798 on Build GRCh38
rs1039954218 Chr.6: 35459803-35459803 on Build GRCh38
rs1038067122 Chr.6: 35459734-35459734 on Build GRCh38
rs185040026 Chr.6: 35459820-35459820 on Build GRCh38
rs1047254399 Chr.6: 35459757-35459758 on Build GRCh38
rs781163272 Chr.6: 35459646-35459646 on Build GRCh38
rs764097845 Chr.6: 35459648-35459648 on Build GRCh38
rs768893669 Chr.6: 35459778-35459778 on Build GRCh38
rs778517461 Chr.6: 35459705-35459705 on Build GRCh38
rs142903218 Chr.6: 35459754-35459754 on Build GRCh38
rs765216017 Chr.6: 35459782-35459782 on Build GRCh38
rs746770705 Chr.6: 35459753-35459753 on Build GRCh38
rs750761652 Chr.6: 35459661-35459661 on Build GRCh38
rs765940689 Chr.6: 35459672-35459672 on Build GRCh38
rs754599731 Chr.6: 35459704-35459704 on Build GRCh38
rs764652720 Chr.6: 35459720-35459720 on Build GRCh38
rs746340447 Chr.6: 35459690-35459693 on Build GRCh38
rs768050713 Chr.6: 35459655-35459655 on Build GRCh38
rs936988373 Chr.6: 35459706-35459706 on Build GRCh38
rs45471595 Chr.6: 35459819-35459819 on Build GRCh38
rs6457823 Chr.6: 35459779-35459779 on Build GRCh38
rs771088063 Chr.6: 35459633-35459633 on Build GRCh38
rs756557647 Chr.6: 35459669-35459669 on Build GRCh38
rs45439495 Chr.6: 35459616-35459616 on Build GRCh38

View Full Product Details