Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87185620-87185885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMTF1
Entrez Gene ID
9988
Gene Name
cyclin D binding myb like transcription factor 1
Gene Aliases
DMP1, DMTF, MRUL, hDMP1
UniGene
Hs.196129
Gene Chromosome Location
Chr.7: 87152361-87196332 on Build GRCh38
Transcripts
NM_001142326.1 NM_001142327.1 NM_021145.3 NR_024549.1 NR_024550.1 XM_011516735.2 XM_011516737.1 XM_011516739.2 XM_017012865.1 XM_017012866.1 XM_017012867.1 XM_017012868.1 XM_017012869.1 XM_017012870.1 XM_017012871.1 XM_017012872.1 XM_017012873.1 XM_017012874.1 XM_017012875.1 XM_017012876.1 XM_017012877.1 XM_017012878.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6645718 c.1050-1G>T p.? Chr.7: 87185828-87185828 on Build GRCh38
6504652 c.1067T>C p.V356A Chr.7: 87185846-87185846 on Build GRCh38
4885353 c.1093G>A p.D365N Chr.7: 87185872-87185872 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746631439 Chr.7: 87185714-87185714 on Build GRCh38
rs760026427 Chr.7: 87185796-87185796 on Build GRCh38
rs531853076 Chr.7: 87185807-87185807 on Build GRCh38
rs546429392 Chr.7: 87185753-87185753 on Build GRCh38
rs757811452 Chr.7: 87185790-87185794 on Build GRCh38
rs753963419 Chr.7: 87185825-87185825 on Build GRCh38
rs764171250 Chr.7: 87185811-87185811 on Build GRCh38
rs765355242 Chr.7: 87185847-87185847 on Build GRCh38
rs1003186036 Chr.7: 87185656-87185656 on Build GRCh38
rs973430828 Chr.7: 87185775-87185775 on Build GRCh38
rs748692186 Chr.7: 87185783-87185783 on Build GRCh38
rs755419283 Chr.7: 87185859-87185859 on Build GRCh38
rs751604357 Chr.7: 87185848-87185848 on Build GRCh38
rs564488331 Chr.7: 87185786-87185786 on Build GRCh38
rs189287223 Chr.7: 87185692-87185692 on Build GRCh38
rs952075653 Chr.7: 87185830-87185830 on Build GRCh38
rs772470232 Chr.7: 87185806-87185806 on Build GRCh38
rs774791595 Chr.7: 87185791-87185791 on Build GRCh38
rs761730060 Chr.7: 87185835-87185835 on Build GRCh38
rs549983096 Chr.7: 87185810-87185810 on Build GRCh38
rs772802785 Chr.7: 87185782-87185782 on Build GRCh38
rs1019658468 Chr.7: 87185708-87185708 on Build GRCh38
rs961069586 Chr.7: 87185769-87185769 on Build GRCh38

View Full Product Details