Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31546016-31546289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1G2
Entrez Gene ID
534
Gene Name
ATPase H+ transporting V1 subunit G2
Gene Aliases
ATP6G, ATP6G2, NG38, VMA10
UniGene
Hs.249227
Gene Chromosome Location
Chr.6: 31544451-31546848 on Build GRCh38
Transcripts
NM_001204078.1 NM_130463.3 NM_138282.2 NR_037853.1

Gene Symbol
ATP6V1G2-DDX39B
Entrez Gene ID
100532737
Gene Name
ATP6V1G2-DDX39B readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.254042
Gene Chromosome Location
Chr.6: 31530219-31546848 on Build GRCh38
Transcripts
NM_001204078.1 NM_130463.3 NM_138282.2 NR_037853.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
274097 c.142C>T p.R48C Chr.6: 31546150-31546150 on Build GRCh38
358020 c.171C>T p.S57S Chr.6: 31546121-31546121 on Build GRCh38
3624563 c.160G>A p.E54K Chr.6: 31546132-31546132 on Build GRCh38
3624564 c.132G>A p.V44V Chr.6: 31546160-31546160 on Build GRCh38
3861686 c.156G>T p.E52D Chr.6: 31546136-31546136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769732049 Chr.6: 31546091-31546091 on Build GRCh38
rs749225629 Chr.6: 31546193-31546193 on Build GRCh38
rs765617580 Chr.6: 31546232-31546232 on Build GRCh38
rs767004486 Chr.6: 31546133-31546133 on Build GRCh38
rs200909248 Chr.6: 31546198-31546198 on Build GRCh38
rs774025595 Chr.6: 31546097-31546097 on Build GRCh38
rs2523502 Chr.6: 31546087-31546087 on Build GRCh38
rs367650140 Chr.6: 31546260-31546260 on Build GRCh38
rs971498686 Chr.6: 31546261-31546261 on Build GRCh38
rs201901212 Chr.6: 31546170-31546170 on Build GRCh38
rs752782495 Chr.6: 31546108-31546108 on Build GRCh38
rs1012738125 Chr.6: 31546212-31546212 on Build GRCh38
rs772923307 Chr.6: 31546128-31546128 on Build GRCh38
rs1036019212 Chr.6: 31546143-31546143 on Build GRCh38
rs1004199229 Chr.6: 31546191-31546191 on Build GRCh38
rs750951348 Chr.6: 31546226-31546232 on Build GRCh38
rs540258120 Chr.6: 31546066-31546066 on Build GRCh38
rs749107418 Chr.6: 31546092-31546092 on Build GRCh38
rs772790052 Chr.6: 31546209-31546209 on Build GRCh38
rs751656100 Chr.6: 31546158-31546158 on Build GRCh38
rs897522626 Chr.6: 31546166-31546166 on Build GRCh38
rs778632528 Chr.6: 31546200-31546200 on Build GRCh38
rs771524022 Chr.6: 31546225-31546225 on Build GRCh38
rs115288880 Chr.6: 31546081-31546081 on Build GRCh38
rs200015995 Chr.6: 31546134-31546134 on Build GRCh38
rs765015126 Chr.6: 31546061-31546061 on Build GRCh38
rs771855136 Chr.6: 31546202-31546202 on Build GRCh38
rs372009509 Chr.6: 31546186-31546186 on Build GRCh38
rs569440748 Chr.6: 31546140-31546140 on Build GRCh38
rs761196920 Chr.6: 31546132-31546132 on Build GRCh38
rs781666346 Chr.6: 31546222-31546223 on Build GRCh38
rs768154830 Chr.6: 31546094-31546094 on Build GRCh38
rs914965494 Chr.6: 31546234-31546234 on Build GRCh38
rs777244593 Chr.6: 31546226-31546226 on Build GRCh38
rs759160444 Chr.6: 31546213-31546213 on Build GRCh38
rs745804253 Chr.6: 31546090-31546090 on Build GRCh38
rs1004187261 Chr.6: 31546150-31546150 on Build GRCh38
rs777766799 Chr.6: 31546146-31546146 on Build GRCh38
rs759973514 Chr.6: 31546137-31546137 on Build GRCh38
rs373793437 Chr.6: 31546188-31546188 on Build GRCh38
rs750261853 Chr.6: 31546067-31546067 on Build GRCh38
rs771599164 Chr.6: 31546110-31546110 on Build GRCh38
rs746538352 Chr.6: 31546215-31546215 on Build GRCh38
rs761421276 Chr.6: 31546109-31546109 on Build GRCh38
rs756498413 Chr.6: 31546248-31546249 on Build GRCh38
rs958632288 Chr.6: 31546201-31546201 on Build GRCh38
rs758603033 Chr.6: 31546141-31546141 on Build GRCh38
rs760007444 Chr.6: 31546231-31546231 on Build GRCh38

View Full Product Details