Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 225992361-225992604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDE2
Entrez Gene ID
163859
Gene Name
SDE2 telomere maintenance homolog
Gene Aliases
C1orf55, dJ671D7.1
UniGene
Hs.520192
Gene Chromosome Location
Chr.1: 225982702-225999365 on Build GRCh38
Transcripts
NM_152608.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5369336 c.343G>A p.E115K Chr.1: 225992539-225992539 on Build GRCh38
5653824 c.389G>A p.R130Q Chr.1: 225992493-225992493 on Build GRCh38
7123991 c.315A>T p.A105A Chr.1: 225992567-225992567 on Build GRCh38
7123993 c.315-1G>A p.? Chr.1: 225992568-225992568 on Build GRCh38
6240057 c.407G>T p.R136L Chr.1: 225992511-225992511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779504088 Chr.1: 225992400-225992400 on Build GRCh38
rs374145200 Chr.1: 225992535-225992535 on Build GRCh38
rs368528473 Chr.1: 225992502-225992502 on Build GRCh38
rs985549233 Chr.1: 225992572-225992572 on Build GRCh38
rs568129149 Chr.1: 225992493-225992493 on Build GRCh38
rs748693302 Chr.1: 225992491-225992491 on Build GRCh38
rs757096219 Chr.1: 225992451-225992451 on Build GRCh38
rs769925412 Chr.1: 225992511-225992511 on Build GRCh38
rs571007811 Chr.1: 225992441-225992441 on Build GRCh38
rs753343589 Chr.1: 225992560-225992560 on Build GRCh38
rs750113065 Chr.1: 225992543-225992543 on Build GRCh38
rs745421825 Chr.1: 225992383-225992383 on Build GRCh38
rs1011222648 Chr.1: 225992504-225992504 on Build GRCh38
rs201707333 Chr.1: 225992390-225992390 on Build GRCh38
rs200329712 Chr.1: 225992485-225992485 on Build GRCh38
rs751370239 Chr.1: 225992449-225992449 on Build GRCh38
rs767370243 Chr.1: 225992540-225992540 on Build GRCh38
rs776863668 Chr.1: 225992505-225992505 on Build GRCh38
rs755760365 Chr.1: 225992545-225992545 on Build GRCh38
rs752696292 Chr.1: 225992419-225992419 on Build GRCh38
rs375469522 Chr.1: 225992494-225992494 on Build GRCh38
rs913187965 Chr.1: 225992387-225992387 on Build GRCh38
rs746035959 Chr.1: 225992509-225992509 on Build GRCh38
rs755707070 Chr.1: 225992484-225992484 on Build GRCh38
rs771250921 Chr.1: 225992503-225992503 on Build GRCh38
rs771195847 Chr.1: 225992408-225992408 on Build GRCh38
rs374097572 Chr.1: 225992407-225992407 on Build GRCh38
rs765185264 Chr.1: 225992416-225992416 on Build GRCh38
rs552823864 Chr.1: 225992439-225992439 on Build GRCh38
rs908826048 Chr.1: 225992391-225992391 on Build GRCh38
rs776809784 Chr.1: 225992411-225992411 on Build GRCh38
rs534722581 Chr.1: 225992456-225992456 on Build GRCh38
rs766049996 Chr.1: 225992552-225992552 on Build GRCh38
rs535141601 Chr.1: 225992512-225992512 on Build GRCh38
rs759649937 Chr.1: 225992414-225992414 on Build GRCh38
rs184898819 Chr.1: 225992534-225992534 on Build GRCh38
rs531194947 Chr.1: 225992401-225992401 on Build GRCh38
rs772498404 Chr.1: 225992405-225992405 on Build GRCh38
rs750058087 Chr.1: 225992464-225992464 on Build GRCh38
rs376970169 Chr.1: 225992476-225992476 on Build GRCh38
rs199840109 Chr.1: 225992526-225992526 on Build GRCh38
rs761734292 Chr.1: 225992536-225992536 on Build GRCh38

View Full Product Details