Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 58831837-58832082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM63B
Entrez Gene ID
54629
Gene Name
family with sequence similarity 63 member B
Gene Aliases
-
UniGene
Hs.591122
Gene Chromosome Location
Chr.15: 58771192-58861900 on Build GRCh38
Transcripts
NM_001040450.2 NM_001040453.2 XM_011521687.1 XM_011521688.1 XM_011521689.1 XM_011521690.1 XM_017022332.1 XM_017022333.1 XM_017022334.1 XM_017022335.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6227354 c.1317A>G p.E439E Chr.15: 58831865-58831865 on Build GRCh38
1243246 c.1333C>T p.R445W Chr.15: 58831881-58831881 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs763271181 Chr.15: 58831943-58831943 on Build GRCh38
rs775613801 Chr.15: 58831911-58831911 on Build GRCh38
rs548529368 Chr.15: 58832050-58832051 on Build GRCh38
rs770755113 Chr.15: 58831901-58831901 on Build GRCh38
rs751818302 Chr.15: 58831958-58831958 on Build GRCh38
rs767782795 Chr.15: 58831886-58831886 on Build GRCh38
rs749093639 Chr.15: 58831899-58831899 on Build GRCh38
rs762508905 Chr.15: 58831885-58831885 on Build GRCh38
rs937172566 Chr.15: 58831990-58831990 on Build GRCh38
rs534600026 Chr.15: 58831926-58831926 on Build GRCh38
rs186589534 Chr.15: 58831944-58831944 on Build GRCh38
rs777706371 Chr.15: 58831898-58831898 on Build GRCh38
rs761928488 Chr.15: 58831936-58831936 on Build GRCh38
rs577798436 Chr.15: 58831991-58831991 on Build GRCh38
rs887510396 Chr.15: 58832031-58832031 on Build GRCh38
rs1055982058 Chr.15: 58832018-58832018 on Build GRCh38
rs928492549 Chr.15: 58831988-58831988 on Build GRCh38
rs990145019 Chr.15: 58831964-58831964 on Build GRCh38
rs945915246 Chr.15: 58831975-58831975 on Build GRCh38
rs756973050 Chr.15: 58831948-58831949 on Build GRCh38
rs748930746 Chr.15: 58831924-58831925 on Build GRCh38
rs981348282 Chr.15: 58831980-58831980 on Build GRCh38
rs376896450 Chr.15: 58831925-58831925 on Build GRCh38
rs913157268 Chr.15: 58831973-58831973 on Build GRCh38
rs776856734 Chr.15: 58831935-58831935 on Build GRCh38
rs756047364 Chr.15: 58831881-58831881 on Build GRCh38
rs773767131 Chr.15: 58831939-58831939 on Build GRCh38

View Full Product Details