Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132574470-132574725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBRSL1
Entrez Gene ID
57666
Gene Name
fibrosin like 1
Gene Aliases
-
UniGene
Hs.411138
Gene Chromosome Location
Chr.12: 132490440-132585188 on Build GRCh38
Transcripts
NM_001142641.1 XM_005266171.3 XM_005266173.3 XM_005266175.3 XM_005266176.3 XM_005266177.3 XM_011534803.2 XM_011534804.2 XM_011534805.2 XM_011534806.2 XM_011534807.2 XM_011534808.2 XM_011534809.2 XM_011534810.2 XM_011534812.2 XM_011534816.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7331021 c.643G>T p.A215S Chr.12: 132574511-132574511 on Build GRCh38
4629909 c.1764G>A p.P588P Chr.12: 132574498-132574498 on Build GRCh38
4641651 c.1741G>A p.A581T Chr.12: 132574475-132574475 on Build GRCh38
1605827 c.696+9A>G p.? Chr.12: 132574573-132574573 on Build GRCh38
1605826 c.1830+9A>G p.? Chr.12: 132574573-132574573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775818068 Chr.12: 132574641-132574641 on Build GRCh38
rs778424660 Chr.12: 132574552-132574552 on Build GRCh38
rs531135747 Chr.12: 132574692-132574692 on Build GRCh38
rs752003475 Chr.12: 132574659-132574659 on Build GRCh38
rs754887984 Chr.12: 132574546-132574546 on Build GRCh38
rs1026477606 Chr.12: 132574684-132574684 on Build GRCh38
rs937408669 Chr.12: 132574650-132574651 on Build GRCh38
rs370695435 Chr.12: 132574519-132574519 on Build GRCh38
rs758969518 Chr.12: 132574571-132574571 on Build GRCh38
rs747611006 Chr.12: 132574558-132574558 on Build GRCh38
rs1016607379 Chr.12: 132574704-132574704 on Build GRCh38
rs574543264 Chr.12: 132574498-132574498 on Build GRCh38
rs144726929 Chr.12: 132574668-132574668 on Build GRCh38
rs1054498669 Chr.12: 132574606-132574606 on Build GRCh38
rs553541139 Chr.12: 132574564-132574564 on Build GRCh38
rs1000607454 Chr.12: 132574694-132574694 on Build GRCh38
rs764587043 Chr.12: 132574618-132574618 on Build GRCh38
rs371732785 Chr.12: 132574544-132574544 on Build GRCh38
rs924725704 Chr.12: 132574592-132574592 on Build GRCh38
rs771246789 Chr.12: 132574603-132574603 on Build GRCh38
rs751875705 Chr.12: 132574489-132574489 on Build GRCh38
rs544498084 Chr.12: 132574693-132574693 on Build GRCh38
rs1020706004 Chr.12: 132574633-132574633 on Build GRCh38
rs962659837 Chr.12: 132574513-132574513 on Build GRCh38
rs576453383 Chr.12: 132574580-132574580 on Build GRCh38
rs899684054 Chr.12: 132574522-132574522 on Build GRCh38
rs554777082 Chr.12: 132574488-132574488 on Build GRCh38
rs1032161849 Chr.12: 132574597-132574597 on Build GRCh38
rs1000726587 Chr.12: 132574545-132574545 on Build GRCh38
rs186465039 Chr.12: 132574573-132574573 on Build GRCh38
rs927377230 Chr.12: 132574644-132574647 on Build GRCh38
rs545357717 Chr.12: 132574660-132574660 on Build GRCh38
rs1047277625 Chr.12: 132574669-132574669 on Build GRCh38
rs780023111 Chr.12: 132574514-132574514 on Build GRCh38
rs746706811 Chr.12: 132574588-132574588 on Build GRCh38
rs367846633 Chr.12: 132574686-132574686 on Build GRCh38
rs561567809 Chr.12: 132574701-132574701 on Build GRCh38
rs374906256 Chr.12: 132574589-132574589 on Build GRCh38
rs951150700 Chr.12: 132574685-132574685 on Build GRCh38
rs981946290 Chr.12: 132574644-132574645 on Build GRCh38
rs997713539 Chr.12: 132574674-132574674 on Build GRCh38
rs757348153 Chr.12: 132574661-132574661 on Build GRCh38

View Full Product Details