Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14609877-14610136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE3
Entrez Gene ID
84658
Gene Name
adhesion G protein-coupled receptor E3
Gene Aliases
EMR3
UniGene
Hs.658712
Gene Chromosome Location
Chr.19: 14600175-14674918 on Build GRCh38
Transcripts
NM_001204118.1 NM_207390.3 NR_109784.1 NR_109785.1 XM_017026785.1 XM_017026786.1 XM_017026787.1 XM_017026788.1 XM_017026789.1 XM_017026790.1 XM_017026791.1 XM_017026792.1 XM_017026793.1

Gene Symbol
CLEC17A
Entrez Gene ID
388512
Gene Name
C-type lectin domain family 17 member A
Gene Aliases
-
UniGene
Hs.567933
Gene Chromosome Location
Chr.19: 14580777-14612035 on Build GRCh38
Transcripts
NM_001204118.1 NM_207390.3 NR_109784.1 NR_109785.1 XM_017026785.1 XM_017026786.1 XM_017026787.1 XM_017026788.1 XM_017026789.1 XM_017026790.1 XM_017026791.1 XM_017026792.1 XM_017026793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5232270 c.936C>T p.A312A Chr.19: 14610075-14610075 on Build GRCh38
6630150 c.1038C>T p.H346H Chr.19: 14610097-14610097 on Build GRCh38
160033 c.1045G>T p.D349Y Chr.19: 14610104-14610104 on Build GRCh38
3822122 c.1016C>T p.P339L Chr.19: 14610075-14610075 on Build GRCh38
709772 c.1019A>G p.E340G Chr.19: 14610078-14610078 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs979500074 Chr.19: 14609917-14609917 on Build GRCh38
rs1027509037 Chr.19: 14609945-14609945 on Build GRCh38
rs897461567 Chr.19: 14610002-14610002 on Build GRCh38
rs755278132 Chr.19: 14610016-14610016 on Build GRCh38
rs769343011 Chr.19: 14610053-14610053 on Build GRCh38
rs748471703 Chr.19: 14610026-14610026 on Build GRCh38
rs767333885 Chr.19: 14610108-14610108 on Build GRCh38
rs910410803 Chr.19: 14609944-14609944 on Build GRCh38
rs750380113 Chr.19: 14610110-14610110 on Build GRCh38
rs372917327 Chr.19: 14610107-14610107 on Build GRCh38
rs935537288 Chr.19: 14610111-14610111 on Build GRCh38
rs563458946 Chr.19: 14609915-14609915 on Build GRCh38
rs374388185 Chr.19: 14609933-14609933 on Build GRCh38
rs932123349 Chr.19: 14610013-14610013 on Build GRCh38
rs184446719 Chr.19: 14609964-14609964 on Build GRCh38
rs772412745 Chr.19: 14610042-14610042 on Build GRCh38
rs768312458 Chr.19: 14610098-14610098 on Build GRCh38
rs754148819 Chr.19: 14610015-14610015 on Build GRCh38
rs779224647 Chr.19: 14610018-14610018 on Build GRCh38
rs762401802 Chr.19: 14610093-14610093 on Build GRCh38
rs983071111 Chr.19: 14609970-14609970 on Build GRCh38
rs778209513 Chr.19: 14610044-14610044 on Build GRCh38
rs932788354 Chr.19: 14609925-14609925 on Build GRCh38
rs922105268 Chr.19: 14610004-14610004 on Build GRCh38
rs190507275 Chr.19: 14609993-14609993 on Build GRCh38
rs368886798 Chr.19: 14610079-14610079 on Build GRCh38
rs779253046 Chr.19: 14609996-14609996 on Build GRCh38
rs966187367 Chr.19: 14609974-14609974 on Build GRCh38
rs530769417 Chr.19: 14609919-14609919 on Build GRCh38
rs966706928 Chr.19: 14609900-14609900 on Build GRCh38
rs925359191 Chr.19: 14609918-14609918 on Build GRCh38
rs569443282 Chr.19: 14610045-14610045 on Build GRCh38
rs773989623 Chr.19: 14610105-14610105 on Build GRCh38

View Full Product Details