Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 87728235-87728474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSL
Entrez Gene ID
1514
Gene Name
cathepsin L
Gene Aliases
CATL, CTSL1, MEP
UniGene
Hs.731507
Gene Chromosome Location
Chr.9: 87725437-87731469 on Build GRCh38
Transcripts
NM_001257971.1 NM_001257972.1 NM_001257973.1 NM_001912.4 NM_145918.2 XM_005251716.3 XM_011518263.1 XM_017014293.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3659334 c.338A>C p.E113A Chr.9: 87728338-87728338 on Build GRCh38
5998454 c.389A>C p.K130T Chr.9: 87728389-87728389 on Build GRCh38
4614084 c.341delC p.R116fs*11 Chr.9: 87728341-87728341 on Build GRCh38
4675480 c.302G>A p.R101K Chr.9: 87728302-87728302 on Build GRCh38
1110879 c.376G>A p.V126M Chr.9: 87728376-87728376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747231438 Chr.9: 87728404-87728404 on Build GRCh38
rs914752764 Chr.9: 87728419-87728419 on Build GRCh38
rs139868873 Chr.9: 87728351-87728351 on Build GRCh38
rs765975858 Chr.9: 87728375-87728375 on Build GRCh38
rs773084675 Chr.9: 87728329-87728330 on Build GRCh38
rs761730126 Chr.9: 87728359-87728359 on Build GRCh38
rs561888153 Chr.9: 87728412-87728412 on Build GRCh38
rs371209580 Chr.9: 87728403-87728403 on Build GRCh38
rs773791760 Chr.9: 87728312-87728312 on Build GRCh38
rs768288006 Chr.9: 87728259-87728259 on Build GRCh38
rs373367766 Chr.9: 87728413-87728413 on Build GRCh38
rs187160957 Chr.9: 87728328-87728328 on Build GRCh38
rs191495548 Chr.9: 87728440-87728440 on Build GRCh38
rs763345527 Chr.9: 87728318-87728318 on Build GRCh38
rs780766917 Chr.9: 87728292-87728292 on Build GRCh38
rs199854486 Chr.9: 87728418-87728418 on Build GRCh38
rs771328780 Chr.9: 87728433-87728433 on Build GRCh38
rs76801214 Chr.9: 87728350-87728350 on Build GRCh38
rs746964642 Chr.9: 87728380-87728380 on Build GRCh38
rs878921053 Chr.9: 87728346-87728346 on Build GRCh38
rs542217433 Chr.9: 87728374-87728374 on Build GRCh38
rs770016150 Chr.9: 87728307-87728307 on Build GRCh38
rs751087214 Chr.9: 87728376-87728376 on Build GRCh38
rs1033820417 Chr.9: 87728382-87728382 on Build GRCh38
rs746806077 Chr.9: 87728347-87728354 on Build GRCh38
rs758188442 Chr.9: 87728319-87728319 on Build GRCh38
rs373216103 Chr.9: 87728366-87728366 on Build GRCh38
rs975167709 Chr.9: 87728293-87728293 on Build GRCh38
rs377749013 Chr.9: 87728391-87728391 on Build GRCh38
rs995217250 Chr.9: 87728281-87728281 on Build GRCh38
rs770099766 Chr.9: 87728415-87728417 on Build GRCh38
rs778182078 Chr.9: 87728343-87728343 on Build GRCh38

View Full Product Details