Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 148783692-148783933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL1
Entrez Gene ID
8454
Gene Name
cullin 1
Gene Aliases
-
UniGene
Hs.146806
Gene Chromosome Location
Chr.7: 148698178-148801110 on Build GRCh38
Transcripts
NM_003592.2 XM_005250060.4 XM_011516629.2 XM_011516630.2 XM_011516631.2 XM_011516632.2 XM_017012723.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4417400 c.1191+8C>T p.? Chr.7: 148783898-148783898 on Build GRCh38
6502456 c.1114G>C p.D372H Chr.7: 148783813-148783813 on Build GRCh38
7269615 c.1111C>T p.L371F Chr.7: 148783810-148783810 on Build GRCh38
6638032 c.1165G>A p.A389T Chr.7: 148783864-148783864 on Build GRCh38
1448963 c.1122_1123insA p.Y377fs*12 Chr.7: 148783821-148783822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745753972 Chr.7: 148783897-148783897 on Build GRCh38
rs752259434 Chr.7: 148783856-148783856 on Build GRCh38
rs944932773 Chr.7: 148783893-148783893 on Build GRCh38
rs746762540 Chr.7: 148783866-148783866 on Build GRCh38
rs760420266 Chr.7: 148783728-148783728 on Build GRCh38
rs750287721 Chr.7: 148783898-148783899 on Build GRCh38
rs767916548 Chr.7: 148783785-148783785 on Build GRCh38
rs890666706 Chr.7: 148783837-148783837 on Build GRCh38
rs774579406 Chr.7: 148783779-148783779 on Build GRCh38
rs759414657 Chr.7: 148783755-148783755 on Build GRCh38
rs369042023 Chr.7: 148783899-148783899 on Build GRCh38
rs932102179 Chr.7: 148783765-148783765 on Build GRCh38
rs76864767 Chr.7: 148783885-148783885 on Build GRCh38
rs188226657 Chr.7: 148783745-148783745 on Build GRCh38
rs17853287 Chr.7: 148783823-148783823 on Build GRCh38
rs10271133 Chr.7: 148783898-148783898 on Build GRCh38
rs373575432 Chr.7: 148783780-148783780 on Build GRCh38
rs753880174 Chr.7: 148783864-148783864 on Build GRCh38
rs767348666 Chr.7: 148783742-148783748 on Build GRCh38
rs777447474 Chr.7: 148783863-148783863 on Build GRCh38
rs754327387 Chr.7: 148783809-148783809 on Build GRCh38
rs778333921 Chr.7: 148783854-148783854 on Build GRCh38
rs758044206 Chr.7: 148783859-148783859 on Build GRCh38
rs768817272 Chr.7: 148783758-148783758 on Build GRCh38
rs141613772 Chr.7: 148783753-148783753 on Build GRCh38
rs1050574067 Chr.7: 148783773-148783773 on Build GRCh38
rs766675341 Chr.7: 148783806-148783806 on Build GRCh38
rs79484500 Chr.7: 148783880-148783880 on Build GRCh38
rs994348862 Chr.7: 148783807-148783807 on Build GRCh38
rs755440894 Chr.7: 148783847-148783847 on Build GRCh38
rs750720092 Chr.7: 148783794-148783794 on Build GRCh38
rs531504939 Chr.7: 148783797-148783797 on Build GRCh38

View Full Product Details