Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2705624-2705859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBF4
Entrez Gene ID
57593
Gene Name
early B-cell factor 4
Gene Aliases
COE4, O/E-4
UniGene
Hs.471955
Gene Chromosome Location
Chr.20: 2692878-2760108 on Build GRCh38
Transcripts
NM_001110514.1 XM_006723600.1 XM_017027983.1 XM_017027984.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4134381 c.184T>G p.S62A Chr.20: 2705635-2705635 on Build GRCh38
4648386 c.246G>A p.E82E Chr.20: 2705697-2705697 on Build GRCh38
4842383 c.281G>A p.R94Q Chr.20: 2705732-2705732 on Build GRCh38
1410907 c.201C>T p.F67F Chr.20: 2705652-2705652 on Build GRCh38
7305773 c.217G>A p.D73N Chr.20: 2705668-2705668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773599115 Chr.20: 2705668-2705668 on Build GRCh38
rs927158543 Chr.20: 2705706-2705706 on Build GRCh38
rs769908262 Chr.20: 2705773-2705773 on Build GRCh38
rs775299839 Chr.20: 2705647-2705647 on Build GRCh38
rs1013709894 Chr.20: 2705814-2705814 on Build GRCh38
rs995485789 Chr.20: 2705799-2705799 on Build GRCh38
rs760998152 Chr.20: 2705671-2705671 on Build GRCh38
rs905118359 Chr.20: 2705819-2705819 on Build GRCh38
rs751318967 Chr.20: 2705721-2705721 on Build GRCh38
rs764741862 Chr.20: 2705700-2705700 on Build GRCh38
rs766121528 Chr.20: 2705789-2705790 on Build GRCh38
rs758037148 Chr.20: 2705713-2705713 on Build GRCh38
rs1039940746 Chr.20: 2705793-2705793 on Build GRCh38
rs61733891 Chr.20: 2705712-2705712 on Build GRCh38
rs766860031 Chr.20: 2705672-2705672 on Build GRCh38
rs1025628033 Chr.20: 2705811-2705811 on Build GRCh38
rs951450616 Chr.20: 2705812-2705812 on Build GRCh38
rs949119005 Chr.20: 2705732-2705732 on Build GRCh38
rs116885461 Chr.20: 2705731-2705731 on Build GRCh38
rs1007430216 Chr.20: 2705785-2705785 on Build GRCh38
rs757029681 Chr.20: 2705738-2705738 on Build GRCh38
rs535007616 Chr.20: 2705788-2705789 on Build GRCh38
rs758725433 Chr.20: 2705786-2705789 on Build GRCh38
rs940834569 Chr.20: 2705710-2705710 on Build GRCh38
rs377524255 Chr.20: 2705699-2705699 on Build GRCh38
rs754374162 Chr.20: 2705685-2705685 on Build GRCh38
rs539502560 Chr.20: 2705667-2705667 on Build GRCh38
rs745809871 Chr.20: 2705769-2705769 on Build GRCh38
rs781205542 Chr.20: 2705768-2705768 on Build GRCh38
rs1000755833 Chr.20: 2705836-2705836 on Build GRCh38
rs945847650 Chr.20: 2705763-2705763 on Build GRCh38
rs777338481 Chr.20: 2705719-2705719 on Build GRCh38
rs568865352 Chr.20: 2705662-2705662 on Build GRCh38
rs555713835 Chr.20: 2705798-2705798 on Build GRCh38
rs113077292 Chr.20: 2705712-2705846 on Build GRCh38

View Full Product Details