Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 77160784-77161023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNG2
Entrez Gene ID
901
Gene Name
cyclin G2
Gene Aliases
-
UniGene
Hs.740456
Gene Chromosome Location
Chr.4: 77157204-77170060 on Build GRCh38
Transcripts
NM_004354.2 XM_011532398.1 XM_011532399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3606048 c.470C>T p.T157I Chr.4: 77160914-77160914 on Build GRCh38
4337749 c.340_342delGAA p.E115delE Chr.4: 77160784-77160786 on Build GRCh38
1200162 c.349T>C p.C117R Chr.4: 77160793-77160793 on Build GRCh38
6623026 c.418C>T p.R140W Chr.4: 77160862-77160862 on Build GRCh38
3940952 c.380G>A p.R127Q Chr.4: 77160824-77160824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753599403 Chr.4: 77160991-77160991 on Build GRCh38
rs888458609 Chr.4: 77160924-77160924 on Build GRCh38
rs755910490 Chr.4: 77160823-77160823 on Build GRCh38
rs761414649 Chr.4: 77160956-77160956 on Build GRCh38
rs748430206 Chr.4: 77160839-77160839 on Build GRCh38
rs142238141 Chr.4: 77160963-77160963 on Build GRCh38
rs553637461 Chr.4: 77160856-77160856 on Build GRCh38
rs771154292 Chr.4: 77160816-77160817 on Build GRCh38
rs770869006 Chr.4: 77160863-77160863 on Build GRCh38
rs35524230 Chr.4: 77160852-77160852 on Build GRCh38
rs779769252 Chr.4: 77160826-77160826 on Build GRCh38
rs760060047 Chr.4: 77160874-77160874 on Build GRCh38
rs4150063 Chr.4: 77160933-77160933 on Build GRCh38
rs371046527 Chr.4: 77160903-77160903 on Build GRCh38
rs146074949 Chr.4: 77160889-77160889 on Build GRCh38
rs778579768 Chr.4: 77160877-77160877 on Build GRCh38
rs368201582 Chr.4: 77160987-77160987 on Build GRCh38
rs1030241549 Chr.4: 77160940-77160940 on Build GRCh38
rs769589284 Chr.4: 77160896-77160896 on Build GRCh38
rs1016707277 Chr.4: 77160893-77160893 on Build GRCh38
rs750376564 Chr.4: 77160964-77160964 on Build GRCh38
rs749212577 Chr.4: 77160943-77160943 on Build GRCh38
rs756035425 Chr.4: 77160970-77160970 on Build GRCh38
rs777310427 Chr.4: 77160868-77160868 on Build GRCh38
rs1033522669 Chr.4: 77160824-77160824 on Build GRCh38
rs199590151 Chr.4: 77160926-77160926 on Build GRCh38
rs985719287 Chr.4: 77160946-77160946 on Build GRCh38
rs772339004 Chr.4: 77160841-77160841 on Build GRCh38
rs200734239 Chr.4: 77160944-77160944 on Build GRCh38
rs750317357 Chr.4: 77160810-77160810 on Build GRCh38

View Full Product Details