Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 74727177-74727448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FA2H
Entrez Gene ID
79152
Gene Name
fatty acid 2-hydroxylase
Gene Aliases
FAAH, FAH1, FAXDC1, SCS7, SPG35
UniGene
Hs.461329
Gene Chromosome Location
Chr.16: 74712955-74774831 on Build GRCh38
Transcripts
NM_024306.4 XM_011523317.2 XM_011523319.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6002069 c.460C>T p.R154C Chr.16: 74727290-74727290 on Build GRCh38
1284801 c.437A>C p.H146P Chr.16: 74727313-74727313 on Build GRCh38
5530745 c.413G>A p.G138E Chr.16: 74727337-74727337 on Build GRCh38
6260845 c.405C>A p.G135G Chr.16: 74727345-74727345 on Build GRCh38
6195228 c.399G>C p.Q133H Chr.16: 74727351-74727351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953982315 Chr.16: 74727325-74727325 on Build GRCh38
rs936626466 Chr.16: 74727376-74727376 on Build GRCh38
rs767257792 Chr.16: 74727395-74727395 on Build GRCh38
rs372350326 Chr.16: 74727307-74727307 on Build GRCh38
rs761723049 Chr.16: 74727343-74727343 on Build GRCh38
rs561163080 Chr.16: 74727363-74727363 on Build GRCh38
rs779450001 Chr.16: 74727214-74727214 on Build GRCh38
rs387907040 Chr.16: 74727290-74727290 on Build GRCh38
rs775608310 Chr.16: 74727270-74727270 on Build GRCh38
rs748854992 Chr.16: 74727212-74727212 on Build GRCh38
rs745430083 Chr.16: 74727263-74727263 on Build GRCh38
rs774231502 Chr.16: 74727287-74727287 on Build GRCh38
rs199659429 Chr.16: 74727349-74727349 on Build GRCh38
rs764721748 Chr.16: 74727375-74727375 on Build GRCh38
rs1003907559 Chr.16: 74727235-74727235 on Build GRCh38
rs757737719 Chr.16: 74727384-74727384 on Build GRCh38
rs763314788 Chr.16: 74727351-74727351 on Build GRCh38
rs149561762 Chr.16: 74727274-74727274 on Build GRCh38
rs751867997 Chr.16: 74727383-74727383 on Build GRCh38
rs749140634 Chr.16: 74727339-74727339 on Build GRCh38
rs926765011 Chr.16: 74727330-74727330 on Build GRCh38
rs770624203 Chr.16: 74727260-74727260 on Build GRCh38
rs748842553 Chr.16: 74727416-74727416 on Build GRCh38
rs758742229 Chr.16: 74727246-74727246 on Build GRCh38
rs886052290 Chr.16: 74727321-74727321 on Build GRCh38
rs755862883 Chr.16: 74727411-74727411 on Build GRCh38
rs142676543 Chr.16: 74727324-74727324 on Build GRCh38
rs867338122 Chr.16: 74727370-74727370 on Build GRCh38
rs779362846 Chr.16: 74727412-74727412 on Build GRCh38
rs746605230 Chr.16: 74727253-74727253 on Build GRCh38
rs777658535 Chr.16: 74727349-74727349 on Build GRCh38
rs751969460 Chr.16: 74727317-74727317 on Build GRCh38
rs925293378 Chr.16: 74727366-74727366 on Build GRCh38
rs373685646 Chr.16: 74727207-74727207 on Build GRCh38
rs201250822 Chr.16: 74727327-74727327 on Build GRCh38
rs777935324 Chr.16: 74727315-74727315 on Build GRCh38
rs201162733 Chr.16: 74727313-74727313 on Build GRCh38
rs138087787 Chr.16: 74727306-74727306 on Build GRCh38
rs753898846 Chr.16: 74727299-74727299 on Build GRCh38
rs199761924 Chr.16: 74727211-74727211 on Build GRCh38
rs771134202 Chr.16: 74727344-74727344 on Build GRCh38
rs766599125 Chr.16: 74727294-74727294 on Build GRCh38
rs138240770 Chr.16: 74727271-74727271 on Build GRCh38
rs569006591 Chr.16: 74727424-74727424 on Build GRCh38
rs765264675 Chr.16: 74727346-74727346 on Build GRCh38
rs371859188 Chr.16: 74727264-74727264 on Build GRCh38
rs760973974 Chr.16: 74727199-74727199 on Build GRCh38
rs769905089 Chr.16: 74727231-74727231 on Build GRCh38
rs531719310 Chr.16: 74727390-74727390 on Build GRCh38
rs986558962 Chr.16: 74727355-74727355 on Build GRCh38
rs141515634 Chr.16: 74727408-74727408 on Build GRCh38
rs776875732 Chr.16: 74727345-74727345 on Build GRCh38
rs773958471 Chr.16: 74727340-74727340 on Build GRCh38
rs146187584 Chr.16: 74727425-74727425 on Build GRCh38
rs769477853 Chr.16: 74727326-74727326 on Build GRCh38
rs146060468 Chr.16: 74727279-74727279 on Build GRCh38
rs753097023 Chr.16: 74727243-74727243 on Build GRCh38
rs781059993 Chr.16: 74727320-74727320 on Build GRCh38
rs901539596 Chr.16: 74727336-74727336 on Build GRCh38
rs778226239 Chr.16: 74727247-74727247 on Build GRCh38
rs886052291 Chr.16: 74727365-74727365 on Build GRCh38

View Full Product Details