Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73410223-73410478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALB
Entrez Gene ID
213
Gene Name
albumin
Gene Aliases
ANALBA, FDAH, PRO0883, PRO0903, PRO1341
UniGene
Hs.418167
Gene Chromosome Location
Chr.4: 73404255-73421412 on Build GRCh38
Transcripts
NM_000477.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6204043 c.659_660delAG p.R221fs*30 Chr.4: 73410355-73410356 on Build GRCh38
6598128 c.653C>T p.A218V Chr.4: 73410349-73410349 on Build GRCh38
4432772 c.618C>T p.L206L Chr.4: 73410314-73410314 on Build GRCh38
1633766 c.667delA p.K223fs*18 Chr.4: 73410363-73410363 on Build GRCh38
1633767 c.667A>C p.K223Q Chr.4: 73410363-73410363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756946865 Chr.4: 73410284-73410284 on Build GRCh38
rs987708371 Chr.4: 73410312-73410312 on Build GRCh38
rs748033134 Chr.4: 73410434-73410434 on Build GRCh38
rs3204504 Chr.4: 73410340-73410340 on Build GRCh38
rs11538220 Chr.4: 73410336-73410336 on Build GRCh38
rs754973519 Chr.4: 73410432-73410432 on Build GRCh38
rs370718057 Chr.4: 73410260-73410260 on Build GRCh38
rs776771984 Chr.4: 73410428-73410434 on Build GRCh38
rs933061735 Chr.4: 73410316-73410316 on Build GRCh38
rs146587652 Chr.4: 73410317-73410317 on Build GRCh38
rs61375018 Chr.4: 73410309-73410309 on Build GRCh38
rs373929336 Chr.4: 73410371-73410371 on Build GRCh38
rs768570250 Chr.4: 73410350-73410353 on Build GRCh38
rs560693428 Chr.4: 73410314-73410314 on Build GRCh38
rs746106868 Chr.4: 73410446-73410446 on Build GRCh38
rs267600248 Chr.4: 73410324-73410324 on Build GRCh38
rs780498508 Chr.4: 73410306-73410306 on Build GRCh38
rs749181890 Chr.4: 73410420-73410420 on Build GRCh38
rs866934964 Chr.4: 73410387-73410387 on Build GRCh38
rs140065129 Chr.4: 73410344-73410344 on Build GRCh38
rs779737113 Chr.4: 73410419-73410419 on Build GRCh38
rs3210154 Chr.4: 73410339-73410339 on Build GRCh38
rs867548179 Chr.4: 73410330-73410330 on Build GRCh38
rs763471006 Chr.4: 73410372-73410372 on Build GRCh38
rs751923567 Chr.4: 73410388-73410388 on Build GRCh38
rs531095851 Chr.4: 73410307-73410307 on Build GRCh38
rs751162736 Chr.4: 73410280-73410280 on Build GRCh38
rs764474235 Chr.4: 73410279-73410279 on Build GRCh38
rs765630261 Chr.4: 73410360-73410360 on Build GRCh38
rs17400586 Chr.4: 73410338-73410338 on Build GRCh38
rs771038647 Chr.4: 73410437-73410437 on Build GRCh38
rs781518359 Chr.4: 73410445-73410445 on Build GRCh38
rs756853717 Chr.4: 73410394-73410394 on Build GRCh38
rs74899956 Chr.4: 73410411-73410411 on Build GRCh38
rs370277478 Chr.4: 73410427-73410427 on Build GRCh38
rs55772354 Chr.4: 73410433-73410433 on Build GRCh38
rs963493317 Chr.4: 73410255-73410255 on Build GRCh38
rs768525028 Chr.4: 73410315-73410315 on Build GRCh38
rs377345537 Chr.4: 73410396-73410396 on Build GRCh38
rs759712202 Chr.4: 73410350-73410350 on Build GRCh38
rs919295208 Chr.4: 73410266-73410266 on Build GRCh38
rs58624704 Chr.4: 73410325-73410325 on Build GRCh38
rs3210163 Chr.4: 73410355-73410355 on Build GRCh38
rs1023446338 Chr.4: 73410251-73410251 on Build GRCh38
rs755614142 Chr.4: 73410407-73410407 on Build GRCh38
rs747134745 Chr.4: 73410328-73410328 on Build GRCh38
rs947166151 Chr.4: 73410380-73410380 on Build GRCh38
rs907701962 Chr.4: 73410326-73410326 on Build GRCh38
rs758600949 Chr.4: 73410272-73410272 on Build GRCh38
rs991349056 Chr.4: 73410248-73410248 on Build GRCh38
rs748077270 Chr.4: 73410442-73410442 on Build GRCh38
rs11538228 Chr.4: 73410349-73410349 on Build GRCh38
rs764382664 Chr.4: 73410377-73410377 on Build GRCh38

View Full Product Details