Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 150221694-150221931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLK2
Entrez Gene ID
166614
Gene Name
doublecortin like kinase 2
Gene Aliases
CL2, CLICK-II, CLICK2, CLIK2, DCAMKL2, DCDC3, DCDC3B, DCK2
UniGene
Hs.591683
Gene Chromosome Location
Chr.4: 150078274-150258563 on Build GRCh38
Transcripts
NM_001040260.3 NM_001040261.4 NR_036614.1 XM_005262785.2 XM_005262786.2 XM_017007832.1 XM_017007833.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5692881 c.1174G>C p.E392Q Chr.4: 150221718-150221718 on Build GRCh38
4628261 c.1212T>C p.N404N Chr.4: 150221756-150221756 on Build GRCh38
1051946 c.1193A>T p.K398M Chr.4: 150221737-150221737 on Build GRCh38
5687960 c.1205C>G p.S402C Chr.4: 150221698-150221698 on Build GRCh38
1051949 c.1212T>A p.N404K Chr.4: 150221756-150221756 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754209312 Chr.4: 150221814-150221814 on Build GRCh38
rs775080091 Chr.4: 150221789-150221789 on Build GRCh38
rs752100061 Chr.4: 150221850-150221850 on Build GRCh38
rs948430123 Chr.4: 150221834-150221834 on Build GRCh38
rs965461014 Chr.4: 150221901-150221901 on Build GRCh38
rs749078554 Chr.4: 150221739-150221739 on Build GRCh38
rs912553036 Chr.4: 150221895-150221895 on Build GRCh38
rs767137443 Chr.4: 150221783-150221783 on Build GRCh38
rs534744184 Chr.4: 150221780-150221780 on Build GRCh38
rs760816548 Chr.4: 150221806-150221815 on Build GRCh38
rs763830860 Chr.4: 150221798-150221798 on Build GRCh38
rs774366531 Chr.4: 150221753-150221753 on Build GRCh38
rs763884604 Chr.4: 150221823-150221824 on Build GRCh38
rs906967314 Chr.4: 150221902-150221902 on Build GRCh38
rs1045779169 Chr.4: 150221855-150221855 on Build GRCh38
rs377155742 Chr.4: 150221826-150221826 on Build GRCh38
rs757667201 Chr.4: 150221819-150221819 on Build GRCh38
rs566095628 Chr.4: 150221751-150221751 on Build GRCh38
rs934371737 Chr.4: 150221904-150221904 on Build GRCh38
rs760458502 Chr.4: 150221795-150221795 on Build GRCh38
rs915584511 Chr.4: 150221797-150221797 on Build GRCh38

View Full Product Details