Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75949086-75949339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOX1
Entrez Gene ID
51
Gene Name
acyl-CoA oxidase 1
Gene Aliases
ACOX, PALMCOX, SCOX
UniGene
Hs.464137
Gene Chromosome Location
Chr.17: 75941507-75979434 on Build GRCh38
Transcripts
NM_001185039.1 NM_004035.6 NM_007292.5 XM_011524868.2 XM_011524869.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1386087 c.1728+2T>G p.? Chr.17: 75949215-75949215 on Build GRCh38
4069734 c.1697G>T p.G566V Chr.17: 75949248-75949248 on Build GRCh38
2742520 c.1712C>T p.A571V Chr.17: 75949233-75949233 on Build GRCh38
2742518 c.1728G>A p.Q576Q Chr.17: 75949217-75949217 on Build GRCh38
361183 c.1614C>G p.L538L Chr.17: 75949331-75949331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759071603 Chr.17: 75949178-75949180 on Build GRCh38
rs994309569 Chr.17: 75949288-75949288 on Build GRCh38
rs777502900 Chr.17: 75949193-75949193 on Build GRCh38
rs752605927 Chr.17: 75949289-75949289 on Build GRCh38
rs559900813 Chr.17: 75949150-75949150 on Build GRCh38
rs889815360 Chr.17: 75949230-75949230 on Build GRCh38
rs768753200 Chr.17: 75949232-75949232 on Build GRCh38
rs548780787 Chr.17: 75949201-75949201 on Build GRCh38
rs374525285 Chr.17: 75949240-75949240 on Build GRCh38
rs773645537 Chr.17: 75949235-75949235 on Build GRCh38
rs748619167 Chr.17: 75949234-75949234 on Build GRCh38
rs929042932 Chr.17: 75949189-75949189 on Build GRCh38
rs8079543 Chr.17: 75949156-75949156 on Build GRCh38
rs568893695 Chr.17: 75949262-75949262 on Build GRCh38
rs945205055 Chr.17: 75949107-75949107 on Build GRCh38
rs747122185 Chr.17: 75949211-75949211 on Build GRCh38
rs910484148 Chr.17: 75949129-75949129 on Build GRCh38
rs767004711 Chr.17: 75949275-75949275 on Build GRCh38
rs746091353 Chr.17: 75949195-75949195 on Build GRCh38
rs763887291 Chr.17: 75949292-75949292 on Build GRCh38
rs766192863 Chr.17: 75949245-75949245 on Build GRCh38
rs774049893 Chr.17: 75949250-75949250 on Build GRCh38
rs189363139 Chr.17: 75949291-75949291 on Build GRCh38
rs184542948 Chr.17: 75949173-75949173 on Build GRCh38
rs758555552 Chr.17: 75949199-75949199 on Build GRCh38
rs771610315 Chr.17: 75949106-75949106 on Build GRCh38
rs199559466 Chr.17: 75949181-75949181 on Build GRCh38
rs1042615378 Chr.17: 75949135-75949135 on Build GRCh38
rs369815943 Chr.17: 75949233-75949233 on Build GRCh38

View Full Product Details