Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31145062-31145295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCHCR1
Entrez Gene ID
54535
Gene Name
coiled-coil alpha-helical rod protein 1
Gene Aliases
C6orf18, HCR, SBP
UniGene
Hs.485075
Gene Chromosome Location
Chr.6: 31142439-31158238 on Build GRCh38
Transcripts
NM_001105563.1 NM_001105564.1 NM_019052.3 XM_011514702.2 XM_011514703.1 XM_011514704.2 XM_011514705.1 XM_017010961.1 XM_017010962.1 XM_017010963.1 XM_017010964.1 XM_017010965.1 XM_017010966.1 XM_017010967.1 XM_017010968.1 XM_017010969.1 XM_017010970.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
741400 c.1557G>A p.L519L Chr.6: 31145218-31145218 on Build GRCh38
5996971 c.1541G>A p.R514H Chr.6: 31145234-31145234 on Build GRCh38
5003132 c.1807_1808insC p.R603fs*51 Chr.6: 31145234-31145235 on Build GRCh38
4532352 c.1877G>A p.G626E Chr.6: 31145073-31145073 on Build GRCh38
7194215 c.1862G>A p.R621Q Chr.6: 31145180-31145180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779839053 Chr.6: 31145218-31145218 on Build GRCh38
rs758053234 Chr.6: 31145253-31145255 on Build GRCh38
rs778373022 Chr.6: 31145083-31145083 on Build GRCh38
rs367616484 Chr.6: 31145161-31145161 on Build GRCh38
rs3094226 Chr.6: 31145122-31145122 on Build GRCh38
rs3094225 Chr.6: 31145275-31145275 on Build GRCh38
rs144588628 Chr.6: 31145175-31145175 on Build GRCh38
rs899284404 Chr.6: 31145146-31145146 on Build GRCh38
rs201470492 Chr.6: 31145085-31145085 on Build GRCh38
rs776900219 Chr.6: 31145180-31145180 on Build GRCh38
rs748634811 Chr.6: 31145171-31145171 on Build GRCh38
rs751505133 Chr.6: 31145123-31145123 on Build GRCh38
rs148460661 Chr.6: 31145181-31145181 on Build GRCh38
rs1022602600 Chr.6: 31145202-31145202 on Build GRCh38
rs754583644 Chr.6: 31145128-31145128 on Build GRCh38
rs184553289 Chr.6: 31145125-31145125 on Build GRCh38
rs762225280 Chr.6: 31145084-31145084 on Build GRCh38
rs773444233 Chr.6: 31145105-31145105 on Build GRCh38
rs777233477 Chr.6: 31145270-31145271 on Build GRCh38
rs771140403 Chr.6: 31145178-31145178 on Build GRCh38
rs567133818 Chr.6: 31145138-31145138 on Build GRCh38
rs751470127 Chr.6: 31145203-31145203 on Build GRCh38
rs778321479 Chr.6: 31145240-31145240 on Build GRCh38
rs748801113 Chr.6: 31145147-31145147 on Build GRCh38
rs552019115 Chr.6: 31145216-31145216 on Build GRCh38
rs747364417 Chr.6: 31145241-31145241 on Build GRCh38
rs754470763 Chr.6: 31145233-31145233 on Build GRCh38
rs752381617 Chr.6: 31145113-31145114 on Build GRCh38
rs2073719 Chr.6: 31145148-31145148 on Build GRCh38
rs201259956 Chr.6: 31145249-31145249 on Build GRCh38
rs373003491 Chr.6: 31145174-31145174 on Build GRCh38
rs759682734 Chr.6: 31145116-31145116 on Build GRCh38
rs557052806 Chr.6: 31145217-31145217 on Build GRCh38
rs765268981 Chr.6: 31145200-31145200 on Build GRCh38
rs776807780 Chr.6: 31145250-31145250 on Build GRCh38
rs575266029 Chr.6: 31145228-31145228 on Build GRCh38
rs779638131 Chr.6: 31145144-31145144 on Build GRCh38
rs766390211 Chr.6: 31145113-31145113 on Build GRCh38
rs750043921 Chr.6: 31145134-31145134 on Build GRCh38
rs199923626 Chr.6: 31145235-31145235 on Build GRCh38
rs3132539 Chr.6: 31145253-31145253 on Build GRCh38
rs760911154 Chr.6: 31145107-31145107 on Build GRCh38
rs188955569 Chr.6: 31145205-31145205 on Build GRCh38
rs761674916 Chr.6: 31145204-31145204 on Build GRCh38
rs753921504 Chr.6: 31145114-31145114 on Build GRCh38
rs748632015 Chr.6: 31145234-31145234 on Build GRCh38

View Full Product Details