Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85107864-85108131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM92B
Entrez Gene ID
339145
Gene Name
family with sequence similarity 92 member B
Gene Aliases
-
UniGene
Hs.125713
Gene Chromosome Location
Chr.16: 85098352-85112508 on Build GRCh38
Transcripts
NM_198491.2 XM_005255931.2 XM_011523063.1 XM_017023198.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3513066 c.376G>A p.E126K Chr.16: 85107896-85107896 on Build GRCh38
4579342 c.360T>C p.H120H Chr.16: 85107912-85107912 on Build GRCh38
4063336 c.309G>T p.Q103H Chr.16: 85108046-85108046 on Build GRCh38
6660343 c.325-7_325-6insT p.? Chr.16: 85107953-85107954 on Build GRCh38
5519413 c.324+8G>A p.? Chr.16: 85108023-85108023 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037408892 Chr.16: 85107946-85107946 on Build GRCh38
rs921975898 Chr.16: 85107987-85107987 on Build GRCh38
rs760598196 Chr.16: 85107973-85107973 on Build GRCh38
rs970532273 Chr.16: 85108062-85108062 on Build GRCh38
rs980583291 Chr.16: 85107910-85107910 on Build GRCh38
rs200258343 Chr.16: 85108110-85108110 on Build GRCh38
rs755342432 Chr.16: 85108069-85108069 on Build GRCh38
rs67587184 Chr.16: 85107950-85107950 on Build GRCh38
rs368660479 Chr.16: 85107957-85107957 on Build GRCh38
rs201885091 Chr.16: 85108076-85108076 on Build GRCh38
rs17200833 Chr.16: 85108055-85108055 on Build GRCh38
rs151242681 Chr.16: 85108099-85108099 on Build GRCh38
rs148587987 Chr.16: 85108032-85108032 on Build GRCh38
rs756627562 Chr.16: 85107994-85107994 on Build GRCh38
rs759415330 Chr.16: 85108024-85108024 on Build GRCh38
rs770931335 Chr.16: 85108081-85108081 on Build GRCh38
rs750138769 Chr.16: 85107885-85107885 on Build GRCh38
rs772190213 Chr.16: 85108007-85108007 on Build GRCh38
rs941917123 Chr.16: 85107944-85107944 on Build GRCh38
rs760642162 Chr.16: 85108013-85108013 on Build GRCh38
rs753740199 Chr.16: 85107976-85107976 on Build GRCh38
rs773226212 Chr.16: 85108010-85108010 on Build GRCh38
rs199676236 Chr.16: 85108098-85108098 on Build GRCh38
rs757927247 Chr.16: 85107984-85107984 on Build GRCh38
rs768905864 Chr.16: 85107965-85107965 on Build GRCh38
rs754145020 Chr.16: 85108068-85108068 on Build GRCh38
rs765000325 Chr.16: 85107980-85107980 on Build GRCh38
rs749522050 Chr.16: 85108080-85108083 on Build GRCh38
rs200208728 Chr.16: 85108109-85108109 on Build GRCh38
rs139476885 Chr.16: 85107888-85107888 on Build GRCh38
rs768489264 Chr.16: 85108062-85108062 on Build GRCh38
rs149250358 Chr.16: 85107897-85107897 on Build GRCh38
rs779316955 Chr.16: 85108112-85108112 on Build GRCh38
rs747077441 Chr.16: 85108077-85108077 on Build GRCh38
rs144820922 Chr.16: 85107898-85107898 on Build GRCh38
rs369301534 Chr.16: 85108023-85108023 on Build GRCh38
rs894192130 Chr.16: 85108082-85108082 on Build GRCh38
rs761327978 Chr.16: 85108106-85108106 on Build GRCh38
rs752194438 Chr.16: 85107981-85107981 on Build GRCh38
rs200655439 Chr.16: 85107967-85107967 on Build GRCh38
rs368454309 Chr.16: 85107972-85107972 on Build GRCh38
rs749489528 Chr.16: 85107964-85107964 on Build GRCh38
rs746832291 Chr.16: 85107964-85107964 on Build GRCh38
rs201188450 Chr.16: 85108097-85108097 on Build GRCh38
rs778547884 Chr.16: 85107923-85107923 on Build GRCh38
rs748392636 Chr.16: 85108073-85108073 on Build GRCh38
rs775489646 Chr.16: 85108033-85108033 on Build GRCh38
rs774571586 Chr.16: 85108006-85108006 on Build GRCh38
rs140072615 Chr.16: 85108064-85108064 on Build GRCh38
rs201498601 Chr.16: 85107939-85107939 on Build GRCh38
rs144351372 Chr.16: 85108039-85108039 on Build GRCh38
rs562032530 Chr.16: 85108086-85108086 on Build GRCh38
rs200618767 Chr.16: 85108003-85108003 on Build GRCh38
rs375723278 Chr.16: 85108014-85108014 on Build GRCh38
rs763635630 Chr.16: 85108048-85108048 on Build GRCh38
rs758443637 Chr.16: 85107886-85107894 on Build GRCh38
rs372330969 Chr.16: 85107966-85107966 on Build GRCh38
rs776711053 Chr.16: 85108083-85108083 on Build GRCh38
rs1036669832 Chr.16: 85108026-85108026 on Build GRCh38
rs370586803 Chr.16: 85107912-85107912 on Build GRCh38
rs978354414 Chr.16: 85107962-85107962 on Build GRCh38
rs762826373 Chr.16: 85108043-85108043 on Build GRCh38
rs199599166 Chr.16: 85108054-85108054 on Build GRCh38
rs780780259 Chr.16: 85108075-85108075 on Build GRCh38
rs772236350 Chr.16: 85108074-85108074 on Build GRCh38
rs143846697 Chr.16: 85107986-85107986 on Build GRCh38
rs746149864 Chr.16: 85107949-85107949 on Build GRCh38
rs768818033 Chr.16: 85108005-85108005 on Build GRCh38
rs267604672 Chr.16: 85107896-85107896 on Build GRCh38
rs781727943 Chr.16: 85107945-85107945 on Build GRCh38
rs545807222 Chr.16: 85107996-85107996 on Build GRCh38
rs753539064 Chr.16: 85107906-85107906 on Build GRCh38
rs769696947 Chr.16: 85108087-85108087 on Build GRCh38
rs766187337 Chr.16: 85107974-85107974 on Build GRCh38
rs199875734 Chr.16: 85108075-85108075 on Build GRCh38
rs754633116 Chr.16: 85107977-85107977 on Build GRCh38
rs780013361 Chr.16: 85107961-85107961 on Build GRCh38

View Full Product Details