Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150997170-150997443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM63A
Entrez Gene ID
55793
Gene Name
family with sequence similarity 63 member A
Gene Aliases
-
UniGene
Hs.743952
Gene Chromosome Location
Chr.1: 150996737-151008378 on Build GRCh38
Transcripts
NM_001040217.2 NM_001163258.1 NM_001163259.1 NM_001163260.1 NM_001319998.1 NM_018379.4 XM_005245321.3 XM_005245325.3 XM_005245327.3 XM_005245328.3 XM_011509757.2 XM_011509759.2 XM_017001770.1 XM_017001771.1 XM_017001772.1 XM_017001773.1 XM_017001774.1 XM_017001775.1 XM_017001776.1 XM_017001777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6260517 c.1398C>A p.C466* Chr.1: 150997299-150997299 on Build GRCh38
895663 c.1330-1G>A p.? Chr.1: 150997368-150997368 on Build GRCh38
3771447 c.1360G>T p.G454W Chr.1: 150997337-150997337 on Build GRCh38
895662 c.1370G>A p.R457Q Chr.1: 150997327-150997327 on Build GRCh38
6260516 c.1542C>A p.C514* Chr.1: 150997299-150997299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776448856 Chr.1: 150997220-150997220 on Build GRCh38
rs754411788 Chr.1: 150997321-150997321 on Build GRCh38
rs765896143 Chr.1: 150997317-150997317 on Build GRCh38
rs587667595 Chr.1: 150997339-150997339 on Build GRCh38
rs774523136 Chr.1: 150997295-150997295 on Build GRCh38
rs587682805 Chr.1: 150997385-150997385 on Build GRCh38
rs587714270 Chr.1: 150997286-150997286 on Build GRCh38
rs772684343 Chr.1: 150997302-150997302 on Build GRCh38
rs911946033 Chr.1: 150997222-150997222 on Build GRCh38
rs775945770 Chr.1: 150997308-150997308 on Build GRCh38
rs745888286 Chr.1: 150997299-150997299 on Build GRCh38
rs781248774 Chr.1: 150997246-150997246 on Build GRCh38
rs770653066 Chr.1: 150997360-150997360 on Build GRCh38
rs374453452 Chr.1: 150997327-150997327 on Build GRCh38
rs752668722 Chr.1: 150997326-150997326 on Build GRCh38
rs528686852 Chr.1: 150997311-150997311 on Build GRCh38
rs587724501 Chr.1: 150997337-150997337 on Build GRCh38
rs754961919 Chr.1: 150997236-150997236 on Build GRCh38
rs755969150 Chr.1: 150997278-150997278 on Build GRCh38
rs759143563 Chr.1: 150997374-150997374 on Build GRCh38
rs773866607 Chr.1: 150997361-150997361 on Build GRCh38
rs762058839 Chr.1: 150997350-150997350 on Build GRCh38
rs770989794 Chr.1: 150997293-150997293 on Build GRCh38
rs747516882 Chr.1: 150997412-150997412 on Build GRCh38
rs202059139 Chr.1: 150997313-150997313 on Build GRCh38
rs778198227 Chr.1: 150997280-150997280 on Build GRCh38
rs1057374052 Chr.1: 150997328-150997328 on Build GRCh38
rs761099204 Chr.1: 150997310-150997310 on Build GRCh38
rs778982251 Chr.1: 150997334-150997334 on Build GRCh38
rs140375829 Chr.1: 150997338-150997338 on Build GRCh38
rs1015017543 Chr.1: 150997384-150997384 on Build GRCh38
rs747991776 Chr.1: 150997269-150997269 on Build GRCh38
rs201983888 Chr.1: 150997346-150997346 on Build GRCh38
rs370021640 Chr.1: 150997318-150997318 on Build GRCh38
rs757524284 Chr.1: 150997331-150997331 on Build GRCh38
rs587622796 Chr.1: 150997375-150997375 on Build GRCh38
rs947534164 Chr.1: 150997219-150997219 on Build GRCh38
rs866033020 Chr.1: 150997343-150997343 on Build GRCh38
rs779194906 Chr.1: 150997248-150997248 on Build GRCh38
rs767625772 Chr.1: 150997325-150997325 on Build GRCh38

View Full Product Details