Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 131275870-131276111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM78A
Entrez Gene ID
286336
Gene Name
family with sequence similarity 78 member A
Gene Aliases
C9orf59
UniGene
Hs.143878
Gene Chromosome Location
Chr.9: 131258078-131281930 on Build GRCh38
Transcripts
NM_033387.3 XM_011518568.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6348026 c.282C>A p.C94* Chr.9: 131275898-131275898 on Build GRCh38
1106098 c.188G>A p.R63H Chr.9: 131275992-131275992 on Build GRCh38
4391533 c.273C>T p.I91I Chr.9: 131275907-131275907 on Build GRCh38
1106097 c.277G>A p.A93T Chr.9: 131275903-131275903 on Build GRCh38
4456137 c.204_205CT>TC p.F69L Chr.9: 131275975-131275976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748456816 Chr.9: 131275911-131275911 on Build GRCh38
rs778086225 Chr.9: 131276028-131276028 on Build GRCh38
rs747791902 Chr.9: 131276007-131276007 on Build GRCh38
rs779384438 Chr.9: 131276077-131276077 on Build GRCh38
rs144598049 Chr.9: 131276009-131276009 on Build GRCh38
rs888932021 Chr.9: 131275934-131275934 on Build GRCh38
rs562235569 Chr.9: 131276062-131276062 on Build GRCh38
rs755408405 Chr.9: 131276022-131276026 on Build GRCh38
rs776522545 Chr.9: 131275942-131275942 on Build GRCh38
rs780076193 Chr.9: 131275986-131275986 on Build GRCh38
rs773630665 Chr.9: 131275915-131275915 on Build GRCh38
rs147422434 Chr.9: 131275964-131275964 on Build GRCh38
rs1054293283 Chr.9: 131275925-131275925 on Build GRCh38
rs867596309 Chr.9: 131275954-131275954 on Build GRCh38
rs768156958 Chr.9: 131275895-131275895 on Build GRCh38
rs545925341 Chr.9: 131275992-131275992 on Build GRCh38
rs763245626 Chr.9: 131276079-131276079 on Build GRCh38
rs750293069 Chr.9: 131275965-131275965 on Build GRCh38
rs748856582 Chr.9: 131275893-131275893 on Build GRCh38
rs754437700 Chr.9: 131275920-131275923 on Build GRCh38
rs769914960 Chr.9: 131276064-131276064 on Build GRCh38
rs188850041 Chr.9: 131275901-131275901 on Build GRCh38
rs759692090 Chr.9: 131275946-131275946 on Build GRCh38
rs915607693 Chr.9: 131275957-131275957 on Build GRCh38
rs766916211 Chr.9: 131275937-131275937 on Build GRCh38
rs765307830 Chr.9: 131275947-131275947 on Build GRCh38
rs772409320 Chr.9: 131275912-131275912 on Build GRCh38
rs752976320 Chr.9: 131275952-131275952 on Build GRCh38
rs774060213 Chr.9: 131275904-131275904 on Build GRCh38
rs775755133 Chr.9: 131276070-131276070 on Build GRCh38
rs140751265 Chr.9: 131275994-131275994 on Build GRCh38
rs543936294 Chr.9: 131276041-131276041 on Build GRCh38
rs758761500 Chr.9: 131275955-131275955 on Build GRCh38
rs747395501 Chr.9: 131276033-131276033 on Build GRCh38
rs772003150 Chr.9: 131275907-131275907 on Build GRCh38
rs377297897 Chr.9: 131275935-131275935 on Build GRCh38
rs756071317 Chr.9: 131275979-131275979 on Build GRCh38

View Full Product Details