Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34792015-34792266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTC1
Entrez Gene ID
70
Gene Name
actin, alpha, cardiac muscle 1
Gene Aliases
ACTC, ASD5, CMD1R, CMH11, LVNC4
UniGene
Hs.118127
Gene Chromosome Location
Chr.15: 34788096-34795726 on Build GRCh38
Transcripts
NM_005159.4 NR_120329.1

Gene Symbol
LOC101928174
Entrez Gene ID
101928174
Gene Name
uncharacterized LOC101928174
Gene Aliases
-
UniGene
Hs.680670
Gene Chromosome Location
Chr.15: 34755084-34812923 on Build GRCh38
Transcripts
NM_005159.4 NR_120329.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4620018 c.772C>T p.R258C Chr.15: 34792126-34792126 on Build GRCh38
2268298 c.752C>G p.T251S Chr.15: 34792146-34792146 on Build GRCh38
3500638 c.727G>A p.E243K Chr.15: 34792171-34792171 on Build GRCh38
2268299 c.740G>A p.G247D Chr.15: 34792158-34792158 on Build GRCh38
6008031 c.701C>A p.S234Y Chr.15: 34792197-34792197 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs397517070 Chr.15: 34792087-34792087 on Build GRCh38
rs1002294264 Chr.15: 34792125-34792125 on Build GRCh38
rs745939876 Chr.15: 34792061-34792061 on Build GRCh38
rs371940910 Chr.15: 34792142-34792142 on Build GRCh38
rs779004124 Chr.15: 34792060-34792060 on Build GRCh38
rs730880399 Chr.15: 34792129-34792129 on Build GRCh38
rs397517068 Chr.15: 34792100-34792100 on Build GRCh38
rs757146524 Chr.15: 34792051-34792051 on Build GRCh38
rs730880396 Chr.15: 34792203-34792203 on Build GRCh38
rs758606996 Chr.15: 34792067-34792067 on Build GRCh38
rs369019849 Chr.15: 34792181-34792181 on Build GRCh38
rs1033276150 Chr.15: 34792216-34792216 on Build GRCh38
rs397517071 Chr.15: 34792092-34792092 on Build GRCh38
rs199566566 Chr.15: 34792122-34792122 on Build GRCh38
rs730880400 Chr.15: 34792089-34792089 on Build GRCh38
rs777933415 Chr.15: 34792060-34792060 on Build GRCh38
rs727505330 Chr.15: 34792159-34792159 on Build GRCh38
rs759078213 Chr.15: 34792220-34792220 on Build GRCh38
rs730880039 Chr.15: 34792147-34792147 on Build GRCh38
rs770397773 Chr.15: 34792185-34792185 on Build GRCh38
rs903582519 Chr.15: 34792124-34792124 on Build GRCh38
rs769158928 Chr.15: 34792106-34792106 on Build GRCh38
rs763817712 Chr.15: 34792047-34792047 on Build GRCh38
rs727504323 Chr.15: 34792105-34792105 on Build GRCh38
rs766868624 Chr.15: 34792235-34792235 on Build GRCh38
rs730880398 Chr.15: 34792143-34792143 on Build GRCh38
rs730880397 Chr.15: 34792198-34792198 on Build GRCh38
rs773788749 Chr.15: 34792210-34792210 on Build GRCh38
rs780321005 Chr.15: 34792071-34792071 on Build GRCh38
rs376724957 Chr.15: 34792075-34792075 on Build GRCh38
rs35907417 Chr.15: 34792044-34792045 on Build GRCh38

View Full Product Details