Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16939823-16940092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CROCC
Entrez Gene ID
9696
Gene Name
ciliary rootlet coiled-coil, rootletin
Gene Aliases
ROLT
UniGene
Hs.309403
Gene Chromosome Location
Chr.1: 16914082-16972979 on Build GRCh38
Transcripts
NM_014675.4 XM_006711058.3 XM_011542468.1 XM_011542470.2 XM_017002913.1 XM_017002914.1 XM_017002915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6378898 c.1643T>C p.L548P Chr.1: 16939928-16939928 on Build GRCh38
1747941 c.1756G>C p.D586H Chr.1: 16940041-16940041 on Build GRCh38
3749010 c.1734C>T p.D578D Chr.1: 16940019-16940019 on Build GRCh38
1336302 c.1621C>T p.R541C Chr.1: 16939906-16939906 on Build GRCh38
677110 c.1630G>T p.A544S Chr.1: 16939915-16939915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779541784 Chr.1: 16939965-16939968 on Build GRCh38
rs773756775 Chr.1: 16940008-16940008 on Build GRCh38
rs758361077 Chr.1: 16940020-16940020 on Build GRCh38
rs767731164 Chr.1: 16939861-16939861 on Build GRCh38
rs774426712 Chr.1: 16939856-16939856 on Build GRCh38
rs371420190 Chr.1: 16939943-16939943 on Build GRCh38
rs201365381 Chr.1: 16940038-16940038 on Build GRCh38
rs780802865 Chr.1: 16939879-16939879 on Build GRCh38
rs774531637 Chr.1: 16939884-16939884 on Build GRCh38
rs141271666 Chr.1: 16940019-16940019 on Build GRCh38
rs572815453 Chr.1: 16939953-16939953 on Build GRCh38
rs550021950 Chr.1: 16939919-16939919 on Build GRCh38
rs762421725 Chr.1: 16939895-16939895 on Build GRCh38
rs369379898 Chr.1: 16940052-16940052 on Build GRCh38
rs748377493 Chr.1: 16940030-16940030 on Build GRCh38
rs759854876 Chr.1: 16939857-16939857 on Build GRCh38
rs778043911 Chr.1: 16940035-16940035 on Build GRCh38
rs758481683 Chr.1: 16939979-16939979 on Build GRCh38
rs145642834 Chr.1: 16939963-16939963 on Build GRCh38
rs79076842 Chr.1: 16939969-16939969 on Build GRCh38
rs755127984 Chr.1: 16940023-16940023 on Build GRCh38
rs367628005 Chr.1: 16939929-16939929 on Build GRCh38
rs371675635 Chr.1: 16939908-16939908 on Build GRCh38
rs147156413 Chr.1: 16939926-16939926 on Build GRCh38
rs770895791 Chr.1: 16939883-16939883 on Build GRCh38
rs558915752 Chr.1: 16940018-16940018 on Build GRCh38
rs775676010 Chr.1: 16939939-16939939 on Build GRCh38
rs565974790 Chr.1: 16939879-16939879 on Build GRCh38
rs145876741 Chr.1: 16939951-16939951 on Build GRCh38
rs192529738 Chr.1: 16939956-16939956 on Build GRCh38
rs759626853 Chr.1: 16939885-16939885 on Build GRCh38
rs374771140 Chr.1: 16939968-16939968 on Build GRCh38
rs750206062 Chr.1: 16940069-16940069 on Build GRCh38
rs753283463 Chr.1: 16940018-16940018 on Build GRCh38
rs201476640 Chr.1: 16940006-16940006 on Build GRCh38
rs371835878 Chr.1: 16939860-16939861 on Build GRCh38
rs752464577 Chr.1: 16939915-16939915 on Build GRCh38
rs769787859 Chr.1: 16939880-16939880 on Build GRCh38
rs374120957 Chr.1: 16939920-16939920 on Build GRCh38
rs780146893 Chr.1: 16939984-16939984 on Build GRCh38
rs781473204 Chr.1: 16940026-16940026 on Build GRCh38
rs761411085 Chr.1: 16940058-16940058 on Build GRCh38
rs760255950 Chr.1: 16940044-16940044 on Build GRCh38
rs751139332 Chr.1: 16939869-16939869 on Build GRCh38
rs776924993 Chr.1: 16940005-16940005 on Build GRCh38
rs763423157 Chr.1: 16940011-16940011 on Build GRCh38
rs375526459 Chr.1: 16940022-16940022 on Build GRCh38
rs768335502 Chr.1: 16940049-16940049 on Build GRCh38
rs767108739 Chr.1: 16940013-16940013 on Build GRCh38
rs145068858 Chr.1: 16940016-16940016 on Build GRCh38
rs751400679 Chr.1: 16939855-16939861 on Build GRCh38
rs749300126 Chr.1: 16939882-16939882 on Build GRCh38
rs761019549 Chr.1: 16939889-16939889 on Build GRCh38
rs202005267 Chr.1: 16939987-16939987 on Build GRCh38
rs547564644 Chr.1: 16939913-16939913 on Build GRCh38
rs767246285 Chr.1: 16939861-16939861 on Build GRCh38
rs141062841 Chr.1: 16939948-16939948 on Build GRCh38
rs368729227 Chr.1: 16940012-16940012 on Build GRCh38
rs765765128 Chr.1: 16939898-16939898 on Build GRCh38
rs559226555 Chr.1: 16939852-16939852 on Build GRCh38
rs754692789 Chr.1: 16939871-16939871 on Build GRCh38
rs112681315 Chr.1: 16939862-16939862 on Build GRCh38
rs761438465 Chr.1: 16939977-16939977 on Build GRCh38
rs763130003 Chr.1: 16939846-16939846 on Build GRCh38
rs757252191 Chr.1: 16939927-16939927 on Build GRCh38
rs768887270 Chr.1: 16939995-16939995 on Build GRCh38
rs762817767 Chr.1: 16940073-16940073 on Build GRCh38
rs745847309 Chr.1: 16939976-16939976 on Build GRCh38
rs144356400 Chr.1: 16939941-16939941 on Build GRCh38
rs767198553 Chr.1: 16939957-16939957 on Build GRCh38
rs778798134 Chr.1: 16939928-16939928 on Build GRCh38
rs772349519 Chr.1: 16939887-16939887 on Build GRCh38
rs371565159 Chr.1: 16939865-16939865 on Build GRCh38
rs764500883 Chr.1: 16939894-16939894 on Build GRCh38
rs371781662 Chr.1: 16939867-16939867 on Build GRCh38
rs772105807 Chr.1: 16939934-16939934 on Build GRCh38
rs1024085663 Chr.1: 16940074-16940074 on Build GRCh38
rs764066247 Chr.1: 16939853-16939854 on Build GRCh38
rs777008883 Chr.1: 16939947-16939947 on Build GRCh38
rs200142599 Chr.1: 16940017-16940017 on Build GRCh38
rs751030538 Chr.1: 16939878-16939879 on Build GRCh38
rs771112973 Chr.1: 16939851-16939851 on Build GRCh38
rs530639652 Chr.1: 16940059-16940059 on Build GRCh38
rs148395884 Chr.1: 16939900-16939900 on Build GRCh38
rs777852121 Chr.1: 16939881-16939881 on Build GRCh38
rs60146471 Chr.1: 16939888-16939888 on Build GRCh38
rs770176672 Chr.1: 16940031-16940031 on Build GRCh38

View Full Product Details