Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20052288-20052553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TANGO2
Entrez Gene ID
128989
Gene Name
transport and golgi organization 2 homolog
Gene Aliases
C22orf25, MECRCN
UniGene
Hs.474233
Gene Chromosome Location
Chr.22: 20017000-20067164 on Build GRCh38
Transcripts
NM_001283106.2 NM_001283116.2 NM_001283129.2 NM_001283148.2 NM_001283154.2 NM_001283179.2 NM_001283186.2 NM_001283199.2 NM_001283215.2 NM_001283235.2 NM_001283248.2 NM_001322141.1 NM_001322142.1 NM_001322143.1 NM_001322144.1 NM_001322145.1 NM_001322146.1 NM_001322147.1 NM_001322148.1 NM_001322149.1 NM_001322150.1 NM_001322153.1 NM_001322155.1 NM_001322160.1 NM_001322163.1 NM_001322166.1 NM_001322167.1 NM_001322169.1 NM_001322171.1 NM_001322172.1 NM_001322173.1 NM_001322174.1 NM_001322175.1 NM_152906.6 NR_104274.2 NR_104275.2 NR_136206.1 NR_136211.1 NR_136212.1 XM_011529863.1 XM_011529865.1 XM_011529867.1 XM_017028577.1 XM_017028578.1 XM_017028579.1 XM_017028580.1 XM_017028581.1 XM_017028582.1 XM_017028583.1 XM_017028584.1 XM_017028585.1 XM_017028586.1 XM_017028587.1 XM_017028588.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6840180 c.309G>C p.L103L Chr.22: 20052505-20052505 on Build GRCh38
6841938 c.322C>T p.R108C Chr.22: 20052518-20052518 on Build GRCh38
6796624 c.199C>A p.R67S Chr.22: 20052518-20052518 on Build GRCh38
6796625 c.214C>A p.R72S Chr.22: 20052518-20052518 on Build GRCh38
6840178 c.186G>C p.L62L Chr.22: 20052505-20052505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574724295 Chr.22: 20052413-20052413 on Build GRCh38
rs948389415 Chr.22: 20052373-20052373 on Build GRCh38
rs766683042 Chr.22: 20052436-20052436 on Build GRCh38
rs559153953 Chr.22: 20052312-20052312 on Build GRCh38
rs576019980 Chr.22: 20052348-20052348 on Build GRCh38
rs370355366 Chr.22: 20052417-20052417 on Build GRCh38
rs771436651 Chr.22: 20052519-20052519 on Build GRCh38
rs3804046 Chr.22: 20052344-20052344 on Build GRCh38
rs771551338 Chr.22: 20052486-20052486 on Build GRCh38
rs111769492 Chr.22: 20052347-20052347 on Build GRCh38
rs889098944 Chr.22: 20052335-20052335 on Build GRCh38
rs758405376 Chr.22: 20052450-20052450 on Build GRCh38
rs971142285 Chr.22: 20052380-20052380 on Build GRCh38
rs772499178 Chr.22: 20052427-20052427 on Build GRCh38
rs373739467 Chr.22: 20052438-20052438 on Build GRCh38
rs778412144 Chr.22: 20052475-20052475 on Build GRCh38
rs978264583 Chr.22: 20052337-20052337 on Build GRCh38
rs776038591 Chr.22: 20052470-20052470 on Build GRCh38
rs867111338 Chr.22: 20052507-20052507 on Build GRCh38
rs747479351 Chr.22: 20052518-20052518 on Build GRCh38
rs749039931 Chr.22: 20052472-20052472 on Build GRCh38
rs981341612 Chr.22: 20052389-20052389 on Build GRCh38
rs542128840 Chr.22: 20052474-20052474 on Build GRCh38
rs1047559159 Chr.22: 20052437-20052437 on Build GRCh38
rs1002584779 Chr.22: 20052381-20052381 on Build GRCh38
rs540584409 Chr.22: 20052462-20052462 on Build GRCh38
rs142975793 Chr.22: 20052349-20052349 on Build GRCh38
rs367921685 Chr.22: 20052443-20052443 on Build GRCh38
rs900493730 Chr.22: 20052500-20052500 on Build GRCh38
rs754597026 Chr.22: 20052473-20052473 on Build GRCh38
rs777290579 Chr.22: 20052515-20052515 on Build GRCh38
rs756140260 Chr.22: 20052469-20052469 on Build GRCh38
rs747899497 Chr.22: 20052482-20052482 on Build GRCh38
rs753021420 Chr.22: 20052448-20052448 on Build GRCh38
rs764311311 Chr.22: 20052458-20052458 on Build GRCh38
rs777015040 Chr.22: 20052442-20052442 on Build GRCh38
rs569610307 Chr.22: 20052342-20052342 on Build GRCh38
rs553980477 Chr.22: 20052533-20052533 on Build GRCh38
rs756009908 Chr.22: 20052531-20052531 on Build GRCh38
rs765391809 Chr.22: 20052447-20052447 on Build GRCh38
rs1031838025 Chr.22: 20052338-20052338 on Build GRCh38

View Full Product Details