Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 25461993-25462266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA2
Entrez Gene ID
157313
Gene Name
cell division cycle associated 2
Gene Aliases
PPP1R81, Repo-Man
UniGene
Hs.33366
Gene Chromosome Location
Chr.8: 25458569-25511308 on Build GRCh38
Transcripts
NM_001317906.1 NM_001317907.1 NM_152562.3 XM_006716294.2 XM_011544415.1 XM_011544416.2 XM_011544417.2 XM_011544418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4670745 c.265C>T p.R89* Chr.8: 25462086-25462086 on Build GRCh38
4670746 c.317G>A p.R106H Chr.8: 25462138-25462138 on Build GRCh38
1098464 c.239C>A p.S80* Chr.8: 25462060-25462060 on Build GRCh38
5030561 c.267A>G p.R89R Chr.8: 25462088-25462088 on Build GRCh38
1098465 c.242C>T p.S81L Chr.8: 25462063-25462063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774444771 Chr.8: 25462067-25462067 on Build GRCh38
rs867731207 Chr.8: 25462182-25462182 on Build GRCh38
rs745822175 Chr.8: 25462131-25462131 on Build GRCh38
rs559652803 Chr.8: 25462089-25462089 on Build GRCh38
rs1034124998 Chr.8: 25462211-25462211 on Build GRCh38
rs751981080 Chr.8: 25462200-25462200 on Build GRCh38
rs757849264 Chr.8: 25462030-25462030 on Build GRCh38
rs778615488 Chr.8: 25462217-25462217 on Build GRCh38
rs746314277 Chr.8: 25462033-25462033 on Build GRCh38
rs756364866 Chr.8: 25462023-25462023 on Build GRCh38
rs765250611 Chr.8: 25462174-25462174 on Build GRCh38
rs535626384 Chr.8: 25462136-25462136 on Build GRCh38
rs773960353 Chr.8: 25462128-25462128 on Build GRCh38
rs114645573 Chr.8: 25462137-25462137 on Build GRCh38
rs758948745 Chr.8: 25462188-25462188 on Build GRCh38
rs750476164 Chr.8: 25462179-25462179 on Build GRCh38
rs149260399 Chr.8: 25462087-25462087 on Build GRCh38
rs748745545 Chr.8: 25462060-25462060 on Build GRCh38
rs748925461 Chr.8: 25462213-25462213 on Build GRCh38
rs142497699 Chr.8: 25462149-25462149 on Build GRCh38
rs780442120 Chr.8: 25462105-25462105 on Build GRCh38
rs767452120 Chr.8: 25462079-25462079 on Build GRCh38
rs951519699 Chr.8: 25462053-25462053 on Build GRCh38
rs759482774 Chr.8: 25462070-25462070 on Build GRCh38
rs775377218 Chr.8: 25462080-25462080 on Build GRCh38
rs371086853 Chr.8: 25462234-25462234 on Build GRCh38
rs781554956 Chr.8: 25462210-25462210 on Build GRCh38
rs780352351 Chr.8: 25462041-25462041 on Build GRCh38
rs769313423 Chr.8: 25462055-25462055 on Build GRCh38
rs756885470 Chr.8: 25462215-25462215 on Build GRCh38
rs778780871 Chr.8: 25462224-25462224 on Build GRCh38
rs765291826 Chr.8: 25462096-25462096 on Build GRCh38
rs116429520 Chr.8: 25462162-25462162 on Build GRCh38
rs762052435 Chr.8: 25462168-25462168 on Build GRCh38
rs745366533 Chr.8: 25462223-25462223 on Build GRCh38
rs372659376 Chr.8: 25462160-25462160 on Build GRCh38
rs766871081 Chr.8: 25462193-25462193 on Build GRCh38
rs776435949 Chr.8: 25462236-25462236 on Build GRCh38
rs781651793 Chr.8: 25462180-25462180 on Build GRCh38
rs755866658 Chr.8: 25462108-25462108 on Build GRCh38
rs755314359 Chr.8: 25462206-25462206 on Build GRCh38
rs144489436 Chr.8: 25462112-25462112 on Build GRCh38
rs368729942 Chr.8: 25462066-25462066 on Build GRCh38
rs761017188 Chr.8: 25462084-25462084 on Build GRCh38
rs753664463 Chr.8: 25462138-25462138 on Build GRCh38
rs757295556 Chr.8: 25462095-25462095 on Build GRCh38
rs779273138 Chr.8: 25462032-25462032 on Build GRCh38
rs747818198 Chr.8: 25462052-25462052 on Build GRCh38
rs753752614 Chr.8: 25462059-25462061 on Build GRCh38
rs747346434 Chr.8: 25462107-25462107 on Build GRCh38
rs761985612 Chr.8: 25462241-25462241 on Build GRCh38
rs772427799 Chr.8: 25462034-25462034 on Build GRCh38
rs116562172 Chr.8: 25462233-25462233 on Build GRCh38
rs748916097 Chr.8: 25462121-25462121 on Build GRCh38
rs758848985 Chr.8: 25462100-25462100 on Build GRCh38
rs750957761 Chr.8: 25462098-25462098 on Build GRCh38
rs142202004 Chr.8: 25462124-25462124 on Build GRCh38

View Full Product Details