Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42804174-42804447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTD1
Entrez Gene ID
124817
Gene Name
cyclin N-terminal domain containing 1
Gene Aliases
CNTD
UniGene
Hs.12272
Gene Chromosome Location
Chr.17: 42798667-42811587 on Build GRCh38
Transcripts
NM_173478.2 XM_005257043.4 XM_005257044.4 XM_005257045.4 XM_006721690.2 XM_011524310.2 XM_011524311.2 XM_017024171.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
231370 c.253G>A p.V85I Chr.17: 42804232-42804232 on Build GRCh38
4673259 c.386A>G p.Q129R Chr.17: 42804365-42804365 on Build GRCh38
1383462 c.410G>A p.R137Q Chr.17: 42804389-42804389 on Build GRCh38
6044815 c.352_353insA p.N118fs*42 Chr.17: 42804331-42804332 on Build GRCh38
3970082 c.255A>T p.V85V Chr.17: 42804234-42804234 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200098048 Chr.17: 42804247-42804247 on Build GRCh38
rs769656924 Chr.17: 42804227-42804227 on Build GRCh38
rs776486844 Chr.17: 42804222-42804222 on Build GRCh38
rs755571397 Chr.17: 42804267-42804267 on Build GRCh38
rs76857741 Chr.17: 42804259-42804259 on Build GRCh38
rs376115344 Chr.17: 42804232-42804232 on Build GRCh38
rs746931837 Chr.17: 42804330-42804332 on Build GRCh38
rs753159130 Chr.17: 42804277-42804277 on Build GRCh38
rs765788831 Chr.17: 42804273-42804273 on Build GRCh38
rs573823531 Chr.17: 42804298-42804298 on Build GRCh38
rs756806659 Chr.17: 42804290-42804290 on Build GRCh38
rs767776718 Chr.17: 42804245-42804245 on Build GRCh38
rs371057489 Chr.17: 42804309-42804309 on Build GRCh38
rs1040949109 Chr.17: 42804230-42804230 on Build GRCh38
rs770466696 Chr.17: 42804214-42804214 on Build GRCh38
rs776792961 Chr.17: 42804368-42804368 on Build GRCh38
rs745835473 Chr.17: 42804225-42804225 on Build GRCh38
rs780662009 Chr.17: 42804411-42804411 on Build GRCh38
rs772060816 Chr.17: 42804305-42804307 on Build GRCh38
rs763465159 Chr.17: 42804393-42804393 on Build GRCh38
rs756969464 Chr.17: 42804410-42804410 on Build GRCh38
rs899719891 Chr.17: 42804241-42804241 on Build GRCh38
rs773365679 Chr.17: 42804351-42804351 on Build GRCh38
rs765882276 Chr.17: 42804388-42804388 on Build GRCh38
rs963758426 Chr.17: 42804321-42804321 on Build GRCh38
rs999542351 Chr.17: 42804395-42804398 on Build GRCh38
rs760733997 Chr.17: 42804258-42804258 on Build GRCh38
rs760955572 Chr.17: 42804357-42804357 on Build GRCh38
rs766891225 Chr.17: 42804231-42804231 on Build GRCh38
rs1002139851 Chr.17: 42804246-42804246 on Build GRCh38
rs762690897 Chr.17: 42804234-42804234 on Build GRCh38
rs1055540641 Chr.17: 42804242-42804242 on Build GRCh38
rs753249329 Chr.17: 42804389-42804389 on Build GRCh38
rs751808274 Chr.17: 42804396-42804396 on Build GRCh38
rs201805683 Chr.17: 42804418-42804418 on Build GRCh38
rs777631230 Chr.17: 42804285-42804285 on Build GRCh38
rs764528802 Chr.17: 42804395-42804395 on Build GRCh38
rs905186605 Chr.17: 42804233-42804233 on Build GRCh38
rs771018364 Chr.17: 42804366-42804366 on Build GRCh38
rs775342957 Chr.17: 42804318-42804319 on Build GRCh38
rs759664970 Chr.17: 42804370-42804370 on Build GRCh38
rs140694460 Chr.17: 42804280-42804280 on Build GRCh38
rs150100281 Chr.17: 42804320-42804320 on Build GRCh38
rs768435292 Chr.17: 42804346-42804346 on Build GRCh38
rs773452433 Chr.17: 42804253-42804253 on Build GRCh38

View Full Product Details