Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 53156960-53157207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD9
Entrez Gene ID
80205
Gene Name
chromodomain helicase DNA binding protein 9
Gene Aliases
AD013, CHD-9, CReMM, KISH2, PRIC320
UniGene
Hs.59159
Gene Chromosome Location
Chr.16: 53054991-53327502 on Build GRCh38
Transcripts
NM_001308319.1 NM_025134.4 XM_005256168.3 XM_005256169.2 XM_005256172.3 XM_006721281.2 XM_006721283.3 XM_011523347.2 XM_017023718.1 XM_017023719.1 XM_017023720.1 XM_017023721.1 XM_017023722.1 XM_017023723.1 XM_017023724.1 XM_017023725.1 XM_017023728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1201026 c.941A>G p.H314R Chr.16: 53157030-53157030 on Build GRCh38
1378307 c.895A>C p.N299H Chr.16: 53156984-53156984 on Build GRCh38
4061191 c.911A>C p.D304A Chr.16: 53157000-53157000 on Build GRCh38
7124316 c.889C>T p.H297Y Chr.16: 53156978-53156978 on Build GRCh38
1201022 c.1115G>A p.G372D Chr.16: 53157204-53157204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768904662 Chr.16: 53157126-53157126 on Build GRCh38
rs764837487 Chr.16: 53157082-53157082 on Build GRCh38
rs762601868 Chr.16: 53157180-53157180 on Build GRCh38
rs368501690 Chr.16: 53157055-53157055 on Build GRCh38
rs746127217 Chr.16: 53157036-53157036 on Build GRCh38
rs770457802 Chr.16: 53157070-53157070 on Build GRCh38
rs372368284 Chr.16: 53157054-53157054 on Build GRCh38
rs751550507 Chr.16: 53157095-53157095 on Build GRCh38
rs773677621 Chr.16: 53157164-53157164 on Build GRCh38
rs890921265 Chr.16: 53157023-53157023 on Build GRCh38
rs61759475 Chr.16: 53157043-53157043 on Build GRCh38
rs200005447 Chr.16: 53157080-53157080 on Build GRCh38
rs781448144 Chr.16: 53157109-53157109 on Build GRCh38
rs776070328 Chr.16: 53157075-53157075 on Build GRCh38
rs952153760 Chr.16: 53157122-53157122 on Build GRCh38
rs908125044 Chr.16: 53157171-53157171 on Build GRCh38
rs772615275 Chr.16: 53157153-53157153 on Build GRCh38
rs757223079 Chr.16: 53157107-53157107 on Build GRCh38
rs535501040 Chr.16: 53157123-53157123 on Build GRCh38
rs763893803 Chr.16: 53157093-53157093 on Build GRCh38
rs1026250197 Chr.16: 53157144-53157144 on Build GRCh38
rs748675330 Chr.16: 53157138-53157138 on Build GRCh38
rs371390517 Chr.16: 53157031-53157031 on Build GRCh38
rs370289002 Chr.16: 53157129-53157129 on Build GRCh38
rs143930746 Chr.16: 53157040-53157040 on Build GRCh38
rs746932672 Chr.16: 53157027-53157027 on Build GRCh38
rs996239832 Chr.16: 53157060-53157060 on Build GRCh38
rs779207024 Chr.16: 53157136-53157136 on Build GRCh38
rs750583516 Chr.16: 53157114-53157114 on Build GRCh38
rs756210035 Chr.16: 53157118-53157118 on Build GRCh38
rs376197651 Chr.16: 53157083-53157083 on Build GRCh38
rs762238500 Chr.16: 53157069-53157069 on Build GRCh38
rs201124945 Chr.16: 53157035-53157035 on Build GRCh38
rs769049307 Chr.16: 53157048-53157048 on Build GRCh38
rs769335335 Chr.16: 53157179-53157179 on Build GRCh38
rs762716450 Chr.16: 53157092-53157092 on Build GRCh38
rs777638518 Chr.16: 53157018-53157018 on Build GRCh38
rs759035782 Chr.16: 53157172-53157172 on Build GRCh38

View Full Product Details