Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36091995-36092248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADPRHL2
Entrez Gene ID
54936
Gene Name
ADP-ribosylhydrolase like 2
Gene Aliases
ARH3
UniGene
Hs.18021
Gene Chromosome Location
Chr.1: 36088852-36093932 on Build GRCh38
Transcripts
NM_017825.2 XM_011541636.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6350433 c.625G>A p.E209K Chr.1: 36092018-36092018 on Build GRCh38
362743 c.691G>A p.D231N Chr.1: 36092084-36092084 on Build GRCh38
7169383 c.636C>T p.L212L Chr.1: 36092029-36092029 on Build GRCh38
908826 c.684C>T p.S228S Chr.1: 36092077-36092077 on Build GRCh38
7305682 c.701+9G>A p.? Chr.1: 36092103-36092103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757224095 Chr.1: 36092141-36092141 on Build GRCh38
rs146990907 Chr.1: 36092043-36092043 on Build GRCh38
rs578209165 Chr.1: 36092205-36092205 on Build GRCh38
rs201851275 Chr.1: 36092072-36092072 on Build GRCh38
rs568872693 Chr.1: 36092146-36092146 on Build GRCh38
rs774652693 Chr.1: 36092065-36092065 on Build GRCh38
rs907722805 Chr.1: 36092136-36092136 on Build GRCh38
rs71649799 Chr.1: 36092110-36092110 on Build GRCh38
rs764811203 Chr.1: 36092077-36092077 on Build GRCh38
rs981669503 Chr.1: 36092036-36092036 on Build GRCh38
rs375407422 Chr.1: 36092120-36092120 on Build GRCh38
rs775954364 Chr.1: 36092071-36092071 on Build GRCh38
rs551436180 Chr.1: 36092212-36092212 on Build GRCh38
rs951684472 Chr.1: 36092031-36092031 on Build GRCh38
rs61748842 Chr.1: 36092017-36092017 on Build GRCh38
rs758065365 Chr.1: 36092022-36092022 on Build GRCh38
rs1053191648 Chr.1: 36092225-36092225 on Build GRCh38
rs779917237 Chr.1: 36092035-36092035 on Build GRCh38
rs372964784 Chr.1: 36092048-36092048 on Build GRCh38
rs907437387 Chr.1: 36092182-36092182 on Build GRCh38
rs189656035 Chr.1: 36092078-36092078 on Build GRCh38
rs938943407 Chr.1: 36092183-36092183 on Build GRCh38
rs772275464 Chr.1: 36092070-36092070 on Build GRCh38
rs761309833 Chr.1: 36092076-36092076 on Build GRCh38
rs933230184 Chr.1: 36092167-36092167 on Build GRCh38
rs778134868 Chr.1: 36092125-36092125 on Build GRCh38
rs1051627125 Chr.1: 36092168-36092168 on Build GRCh38
rs758387348 Chr.1: 36092092-36092092 on Build GRCh38
rs991558329 Chr.1: 36092062-36092062 on Build GRCh38
rs771199144 Chr.1: 36092056-36092056 on Build GRCh38
rs2236387 Chr.1: 36092018-36092018 on Build GRCh38
rs920041760 Chr.1: 36092165-36092165 on Build GRCh38
rs771452373 Chr.1: 36092188-36092188 on Build GRCh38
rs376220008 Chr.1: 36092051-36092051 on Build GRCh38
rs757385182 Chr.1: 36092162-36092162 on Build GRCh38
rs751373162 Chr.1: 36092119-36092119 on Build GRCh38
rs746397934 Chr.1: 36092069-36092069 on Build GRCh38
rs776325194 Chr.1: 36092073-36092073 on Build GRCh38
rs748273224 Chr.1: 36092040-36092040 on Build GRCh38
rs746321037 Chr.1: 36092144-36092144 on Build GRCh38
rs963026089 Chr.1: 36092013-36092013 on Build GRCh38
rs116577025 Chr.1: 36092160-36092160 on Build GRCh38

View Full Product Details