Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133268082-133268345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM8
Entrez Gene ID
101
Gene Name
ADAM metallopeptidase domain 8
Gene Aliases
CD156, CD156a, MS2
UniGene
Hs.501574
Gene Chromosome Location
Chr.10: 133262416-133276903 on Build GRCh38
Transcripts
NM_001109.4 NM_001164489.1 NM_001164490.1 XM_011539117.2 XM_017015465.1

SNP Detail

SNP ID Chromosome Location
rs11101672 Chr.10: 133268250-133268250 on Build GRCh38
rs552903661 Chr.10: 133268200-133268200 on Build GRCh38
rs769181696 Chr.10: 133268100-133268100 on Build GRCh38
rs568514995 Chr.10: 133268289-133268289 on Build GRCh38
rs180836438 Chr.10: 133268127-133268127 on Build GRCh38
rs900208373 Chr.10: 133268155-133268156 on Build GRCh38
rs373868739 Chr.10: 133268138-133268138 on Build GRCh38
rs957649031 Chr.10: 133268206-133268206 on Build GRCh38
rs981645803 Chr.10: 133268293-133268293 on Build GRCh38
rs149418021 Chr.10: 133268114-133268114 on Build GRCh38
rs368072866 Chr.10: 133268139-133268139 on Build GRCh38
rs957900412 Chr.10: 133268211-133268211 on Build GRCh38
rs752666568 Chr.10: 133268163-133268163 on Build GRCh38
rs999465074 Chr.10: 133268179-133268179 on Build GRCh38
rs911344898 Chr.10: 133268228-133268228 on Build GRCh38
rs997417459 Chr.10: 133268169-133268169 on Build GRCh38
rs964037354 Chr.10: 133268231-133268231 on Build GRCh38
rs983992685 Chr.10: 133268313-133268313 on Build GRCh38
rs781132236 Chr.10: 133268170-133268170 on Build GRCh38
rs540784289 Chr.10: 133268154-133268154 on Build GRCh38
rs752998170 Chr.10: 133268133-133268134 on Build GRCh38
rs929492285 Chr.10: 133268124-133268124 on Build GRCh38
rs532766486 Chr.10: 133268181-133268181 on Build GRCh38
rs548607450 Chr.10: 133268271-133268271 on Build GRCh38
rs563217693 Chr.10: 133268237-133268237 on Build GRCh38
rs753601907 Chr.10: 133268153-133268153 on Build GRCh38
rs976772423 Chr.10: 133268232-133268232 on Build GRCh38
rs866868319 Chr.10: 133268107-133268107 on Build GRCh38
rs922835235 Chr.10: 133268244-133268244 on Build GRCh38
rs757475835 Chr.10: 133268167-133268167 on Build GRCh38
rs558306638 Chr.10: 133268120-133268121 on Build GRCh38
rs1009361309 Chr.10: 133268176-133268176 on Build GRCh38
rs1011900690 Chr.10: 133268145-133268145 on Build GRCh38
rs928861614 Chr.10: 133268301-133268301 on Build GRCh38
rs909999027 Chr.10: 133268302-133268302 on Build GRCh38
rs2995309 Chr.10: 133268180-133268180 on Build GRCh38
rs765308893 Chr.10: 133268158-133268158 on Build GRCh38
rs575741573 Chr.10: 133268115-133268115 on Build GRCh38
rs989938222 Chr.10: 133268226-133268226 on Build GRCh38
rs1032176354 Chr.10: 133268193-133268193 on Build GRCh38
rs983072495 Chr.10: 133268294-133268294 on Build GRCh38

View Full Product Details