Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 9791142-9791401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS7C
Entrez Gene ID
201140
Gene Name
dehydrogenase/reductase 7C
Gene Aliases
SDR32C2
UniGene
Hs.61684
Gene Chromosome Location
Chr.17: 9771434-9791297 on Build GRCh38
Transcripts
NM_001105571.2 NM_001220493.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1387738 c.121G>T p.V41L Chr.17: 9791164-9791164 on Build GRCh38
4071193 c.10A>G p.M4V Chr.17: 9791275-9791275 on Build GRCh38
4071192 c.95A>C p.K32T Chr.17: 9791190-9791190 on Build GRCh38
3524140 c.142G>A p.G48R Chr.17: 9791143-9791143 on Build GRCh38
6195300 c.14C>G p.A5G Chr.17: 9791271-9791271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373961243 Chr.17: 9791283-9791283 on Build GRCh38
rs755320240 Chr.17: 9791167-9791167 on Build GRCh38
rs779838304 Chr.17: 9791381-9791381 on Build GRCh38
rs755444121 Chr.17: 9791284-9791284 on Build GRCh38
rs375493603 Chr.17: 9791213-9791213 on Build GRCh38
rs772925322 Chr.17: 9791217-9791217 on Build GRCh38
rs369426897 Chr.17: 9791315-9791315 on Build GRCh38
rs541105527 Chr.17: 9791296-9791296 on Build GRCh38
rs532103187 Chr.17: 9791164-9791164 on Build GRCh38
rs766432562 Chr.17: 9791280-9791280 on Build GRCh38
rs753108299 Chr.17: 9791298-9791298 on Build GRCh38
rs750537560 Chr.17: 9791266-9791266 on Build GRCh38
rs775205464 Chr.17: 9791246-9791246 on Build GRCh38
rs748716707 Chr.17: 9791226-9791226 on Build GRCh38
rs994626038 Chr.17: 9791196-9791196 on Build GRCh38
rs199512145 Chr.17: 9791233-9791233 on Build GRCh38
rs769558034 Chr.17: 9791208-9791208 on Build GRCh38
rs376176859 Chr.17: 9791201-9791201 on Build GRCh38
rs755871389 Chr.17: 9791303-9791303 on Build GRCh38
rs770382678 Chr.17: 9791313-9791313 on Build GRCh38
rs748926190 Chr.17: 9791312-9791312 on Build GRCh38
rs967159932 Chr.17: 9791371-9791371 on Build GRCh38
rs777545444 Chr.17: 9791304-9791304 on Build GRCh38
rs762078139 Chr.17: 9791258-9791258 on Build GRCh38
rs942034460 Chr.17: 9791302-9791302 on Build GRCh38
rs907007029 Chr.17: 9791269-9791269 on Build GRCh38
rs751306781 Chr.17: 9791293-9791293 on Build GRCh38
rs759481316 Chr.17: 9791237-9791237 on Build GRCh38
rs771770633 Chr.17: 9791240-9791240 on Build GRCh38
rs547509926 Chr.17: 9791234-9791234 on Build GRCh38
rs765438036 Chr.17: 9791261-9791261 on Build GRCh38
rs976206539 Chr.17: 9791376-9791376 on Build GRCh38
rs200734929 Chr.17: 9791335-9791335 on Build GRCh38
rs762919426 Chr.17: 9791275-9791275 on Build GRCh38
rs1001304593 Chr.17: 9791272-9791272 on Build GRCh38
rs781654590 Chr.17: 9791186-9791186 on Build GRCh38
rs769788082 Chr.17: 9791334-9791334 on Build GRCh38
rs766644248 Chr.17: 9791243-9791245 on Build GRCh38

View Full Product Details