Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1879962-1880193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD17A
Entrez Gene ID
81926
Gene Name
abhydrolase domain containing 17A
Gene Aliases
C19orf27, FAM108A1
UniGene
Hs.465542
Gene Chromosome Location
Chr.19: 1876965-1885519 on Build GRCh38
Transcripts
NM_001130111.1 NM_031213.3 XM_011528338.1 XM_017027341.1 XM_017027342.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6237948 c.495C>T p.V165V Chr.19: 1880106-1880106 on Build GRCh38
4075722 c.603C>T p.Y201Y Chr.19: 1879998-1879998 on Build GRCh38
6288746 c.513T>C p.N171N Chr.19: 1880088-1880088 on Build GRCh38
115961 c.593A>C p.Y198S Chr.19: 1880008-1880008 on Build GRCh38
5546331 c.584C>T p.S195F Chr.19: 1880017-1880017 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563816057 Chr.19: 1880130-1880130 on Build GRCh38
rs768889344 Chr.19: 1880125-1880125 on Build GRCh38
rs575907020 Chr.19: 1880056-1880056 on Build GRCh38
rs7260332 Chr.19: 1880018-1880018 on Build GRCh38
rs760209282 Chr.19: 1880146-1880146 on Build GRCh38
rs775407484 Chr.19: 1880013-1880013 on Build GRCh38
rs970508069 Chr.19: 1880025-1880025 on Build GRCh38
rs772233054 Chr.19: 1880009-1880009 on Build GRCh38
rs777774406 Chr.19: 1879987-1879987 on Build GRCh38
rs769986857 Chr.19: 1880156-1880157 on Build GRCh38
rs990723562 Chr.19: 1880007-1880007 on Build GRCh38
rs750179630 Chr.19: 1880161-1880161 on Build GRCh38
rs745964941 Chr.19: 1880005-1880005 on Build GRCh38
rs762117498 Chr.19: 1880034-1880034 on Build GRCh38
rs770246378 Chr.19: 1880135-1880135 on Build GRCh38
rs750982677 Chr.19: 1880041-1880041 on Build GRCh38
rs757073884 Chr.19: 1880085-1880085 on Build GRCh38
rs146705668 Chr.19: 1879986-1879986 on Build GRCh38
rs760943932 Chr.19: 1880021-1880021 on Build GRCh38
rs139072903 Chr.19: 1879998-1879998 on Build GRCh38
rs752266091 Chr.19: 1880143-1880143 on Build GRCh38
rs771001154 Chr.19: 1880004-1880004 on Build GRCh38
rs377727671 Chr.19: 1880128-1880128 on Build GRCh38
rs763351670 Chr.19: 1880042-1880042 on Build GRCh38
rs561025016 Chr.19: 1880126-1880126 on Build GRCh38
rs376776483 Chr.19: 1880054-1880054 on Build GRCh38
rs7248295 Chr.19: 1880008-1880008 on Build GRCh38
rs199570784 Chr.19: 1879988-1879988 on Build GRCh38
rs756947520 Chr.19: 1880158-1880158 on Build GRCh38
rs780188216 Chr.19: 1880110-1880110 on Build GRCh38
rs201443342 Chr.19: 1880010-1880010 on Build GRCh38
rs776996252 Chr.19: 1880031-1880031 on Build GRCh38
rs763586472 Chr.19: 1880138-1880138 on Build GRCh38
rs771889080 Chr.19: 1880097-1880097 on Build GRCh38
rs542398863 Chr.19: 1880121-1880121 on Build GRCh38
rs375711582 Chr.19: 1880150-1880150 on Build GRCh38
rs759678716 Chr.19: 1880018-1880020 on Build GRCh38
rs528105267 Chr.19: 1880159-1880159 on Build GRCh38
rs545677149 Chr.19: 1880052-1880052 on Build GRCh38
rs765695569 Chr.19: 1880038-1880038 on Build GRCh38
rs767115355 Chr.19: 1880050-1880050 on Build GRCh38
rs747035503 Chr.19: 1880116-1880116 on Build GRCh38
rs753703744 Chr.19: 1880061-1880061 on Build GRCh38
rs764235356 Chr.19: 1880026-1880026 on Build GRCh38
rs143714194 Chr.19: 1880166-1880166 on Build GRCh38
rs773437593 Chr.19: 1880137-1880137 on Build GRCh38
rs766726328 Chr.19: 1880141-1880141 on Build GRCh38
rs928673298 Chr.19: 1880093-1880093 on Build GRCh38
rs371164909 Chr.19: 1880149-1880149 on Build GRCh38
rs373030033 Chr.19: 1880094-1880094 on Build GRCh38

View Full Product Details