Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15626610-15626851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDI2
Entrez Gene ID
84301
Gene Name
DNA damage inducible 1 homolog 2
Gene Aliases
-
UniGene
Hs.718857
Gene Chromosome Location
Chr.1: 15617458-15661057 on Build GRCh38
Transcripts
NM_032341.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5993111 c.151G>T p.E51* Chr.1: 15626681-15626681 on Build GRCh38
5479500 c.224G>A p.R75Q Chr.1: 15626754-15626754 on Build GRCh38
2084648 c.150G>A p.A50A Chr.1: 15626680-15626680 on Build GRCh38
1319832 c.170A>G p.N57S Chr.1: 15626700-15626700 on Build GRCh38
3476559 c.230A>T p.K77M Chr.1: 15626760-15626760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs921001720 Chr.1: 15626739-15626739 on Build GRCh38
rs752722490 Chr.1: 15626730-15626730 on Build GRCh38
rs144340720 Chr.1: 15626766-15626766 on Build GRCh38
rs376309082 Chr.1: 15626629-15626629 on Build GRCh38
rs750770128 Chr.1: 15626680-15626680 on Build GRCh38
rs758350022 Chr.1: 15626731-15626731 on Build GRCh38
rs755177299 Chr.1: 15626729-15626729 on Build GRCh38
rs150610844 Chr.1: 15626740-15626740 on Build GRCh38
rs756295097 Chr.1: 15626648-15626650 on Build GRCh38
rs942711022 Chr.1: 15626798-15626798 on Build GRCh38
rs766554026 Chr.1: 15626725-15626725 on Build GRCh38
rs772543586 Chr.1: 15626804-15626804 on Build GRCh38
rs367661237 Chr.1: 15626647-15626647 on Build GRCh38
rs780063054 Chr.1: 15626660-15626662 on Build GRCh38
rs761074524 Chr.1: 15626711-15626711 on Build GRCh38
rs757208866 Chr.1: 15626794-15626794 on Build GRCh38
rs913479339 Chr.1: 15626744-15626744 on Build GRCh38
rs749375601 Chr.1: 15626665-15626665 on Build GRCh38
rs901385150 Chr.1: 15626826-15626826 on Build GRCh38
rs370103023 Chr.1: 15626654-15626654 on Build GRCh38
rs774889431 Chr.1: 15626671-15626671 on Build GRCh38
rs765443704 Chr.1: 15626829-15626829 on Build GRCh38
rs372813942 Chr.1: 15626821-15626821 on Build GRCh38
rs72871771 Chr.1: 15626631-15626631 on Build GRCh38
rs199576400 Chr.1: 15626817-15626817 on Build GRCh38
rs751384510 Chr.1: 15626692-15626692 on Build GRCh38
rs561921842 Chr.1: 15626722-15626722 on Build GRCh38
rs374425147 Chr.1: 15626754-15626754 on Build GRCh38
rs1016620809 Chr.1: 15626635-15626635 on Build GRCh38
rs777917292 Chr.1: 15626732-15626732 on Build GRCh38
rs202121145 Chr.1: 15626778-15626778 on Build GRCh38
rs769348446 Chr.1: 15626784-15626784 on Build GRCh38
rs77782333 Chr.1: 15626728-15626728 on Build GRCh38
rs193147217 Chr.1: 15626813-15626813 on Build GRCh38
rs374078440 Chr.1: 15626644-15626644 on Build GRCh38
rs773745181 Chr.1: 15626810-15626810 on Build GRCh38
rs967176786 Chr.1: 15626650-15626650 on Build GRCh38
rs762325900 Chr.1: 15626672-15626672 on Build GRCh38
rs754214672 Chr.1: 15626820-15626820 on Build GRCh38
rs190145902 Chr.1: 15626676-15626676 on Build GRCh38
rs769311230 Chr.1: 15626669-15626669 on Build GRCh38
rs147524577 Chr.1: 15626761-15626761 on Build GRCh38

View Full Product Details