Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57966471-57966724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf18
Entrez Gene ID
147685
Gene Name
chromosome 19 open reading frame 18
Gene Aliases
-
UniGene
Hs.134209
Gene Chromosome Location
Chr.19: 57958437-57974534 on Build GRCh38
Transcripts
NM_152474.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
170391 c.314C>T p.S105L Chr.19: 57966587-57966587 on Build GRCh38
6365640 c.293T>A p.L98H Chr.19: 57966608-57966608 on Build GRCh38
1177772 c.310A>G p.I104V Chr.19: 57966591-57966591 on Build GRCh38
4667131 c.366G>C p.M122I Chr.19: 57966535-57966535 on Build GRCh38
6086265 c.346G>A p.G116R Chr.19: 57966555-57966555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763808608 Chr.19: 57966648-57966648 on Build GRCh38
rs760777736 Chr.19: 57966615-57966615 on Build GRCh38
rs779745202 Chr.19: 57966680-57966680 on Build GRCh38
rs955499380 Chr.19: 57966567-57966567 on Build GRCh38
rs1055487796 Chr.19: 57966613-57966613 on Build GRCh38
rs575359579 Chr.19: 57966690-57966690 on Build GRCh38
rs373357471 Chr.19: 57966660-57966660 on Build GRCh38
rs780592836 Chr.19: 57966582-57966582 on Build GRCh38
rs189213099 Chr.19: 57966623-57966623 on Build GRCh38
rs751776257 Chr.19: 57966614-57966614 on Build GRCh38
rs769105610 Chr.19: 57966583-57966583 on Build GRCh38
rs767436619 Chr.19: 57966528-57966528 on Build GRCh38
rs375464552 Chr.19: 57966571-57966571 on Build GRCh38
rs754746042 Chr.19: 57966685-57966688 on Build GRCh38
rs765216494 Chr.19: 57966542-57966542 on Build GRCh38
rs935883376 Chr.19: 57966603-57966603 on Build GRCh38
rs372518377 Chr.19: 57966508-57966508 on Build GRCh38
rs180867951 Chr.19: 57966656-57966656 on Build GRCh38
rs112908801 Chr.19: 57966580-57966580 on Build GRCh38
rs755643118 Chr.19: 57966578-57966578 on Build GRCh38
rs756936603 Chr.19: 57966552-57966552 on Build GRCh38
rs774671608 Chr.19: 57966505-57966505 on Build GRCh38
rs762908137 Chr.19: 57966647-57966647 on Build GRCh38
rs773374058 Chr.19: 57966610-57966610 on Build GRCh38
rs1048017685 Chr.19: 57966686-57966686 on Build GRCh38
rs772326799 Chr.19: 57966600-57966600 on Build GRCh38
rs146255633 Chr.19: 57966624-57966624 on Build GRCh38
rs773344124 Chr.19: 57966532-57966532 on Build GRCh38
rs750096083 Chr.19: 57966657-57966657 on Build GRCh38
rs141757302 Chr.19: 57966546-57966546 on Build GRCh38
rs369164495 Chr.19: 57966502-57966502 on Build GRCh38
rs989862465 Chr.19: 57966640-57966641 on Build GRCh38
rs574609587 Chr.19: 57966591-57966591 on Build GRCh38
rs780506728 Chr.19: 57966566-57966566 on Build GRCh38
rs886313408 Chr.19: 57966694-57966694 on Build GRCh38
rs774863435 Chr.19: 57966586-57966586 on Build GRCh38
rs754332624 Chr.19: 57966683-57966683 on Build GRCh38
rs751365502 Chr.19: 57966649-57966649 on Build GRCh38
rs751171059 Chr.19: 57966549-57966549 on Build GRCh38
rs750670987 Chr.19: 57966681-57966682 on Build GRCh38
rs934397149 Chr.19: 57966632-57966632 on Build GRCh38
rs200742590 Chr.19: 57966587-57966587 on Build GRCh38
rs138842416 Chr.19: 57966539-57966539 on Build GRCh38
rs752670588 Chr.19: 57966544-57966544 on Build GRCh38
rs949835799 Chr.19: 57966620-57966620 on Build GRCh38

View Full Product Details