Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34997143-34997368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB5
Entrez Gene ID
25822
Gene Name
DnaJ heat shock protein family (Hsp40) member B5
Gene Aliases
Hsc40
UniGene
Hs.237506
Gene Chromosome Location
Chr.9: 34989369-34998433 on Build GRCh38
Transcripts
NM_001135004.2 NM_001135005.2 NM_012266.5 XM_005251428.3 XM_006716751.2 XM_011517842.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1598255 c.1325G>A p.R442H Chr.9: 34997195-34997195 on Build GRCh38
4678673 c.981T>C p.V327V Chr.9: 34997193-34997193 on Build GRCh38
1598256 c.1324C>T p.R442C Chr.9: 34997194-34997194 on Build GRCh38
4678675 c.1025A>C p.K342T Chr.9: 34997237-34997237 on Build GRCh38
319964 c.953G>A p.R318Q Chr.9: 34997165-34997165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770051452 Chr.9: 34997320-34997320 on Build GRCh38
rs908530318 Chr.9: 34997284-34997284 on Build GRCh38
rs914891315 Chr.9: 34997274-34997274 on Build GRCh38
rs752033759 Chr.9: 34997265-34997265 on Build GRCh38
rs143047567 Chr.9: 34997212-34997212 on Build GRCh38
rs765620962 Chr.9: 34997244-34997244 on Build GRCh38
rs775331441 Chr.9: 34997208-34997208 on Build GRCh38
rs779994915 Chr.9: 34997301-34997301 on Build GRCh38
rs745724497 Chr.9: 34997195-34997195 on Build GRCh38
rs369215213 Chr.9: 34997209-34997209 on Build GRCh38
rs534902052 Chr.9: 34997317-34997317 on Build GRCh38
rs368959542 Chr.9: 34997298-34997298 on Build GRCh38
rs745632299 Chr.9: 34997295-34997295 on Build GRCh38
rs762128897 Chr.9: 34997235-34997235 on Build GRCh38
rs771978603 Chr.9: 34997196-34997196 on Build GRCh38
rs373116154 Chr.9: 34997260-34997260 on Build GRCh38
rs746936238 Chr.9: 34997305-34997305 on Build GRCh38
rs921319578 Chr.9: 34997324-34997324 on Build GRCh38
rs773698287 Chr.9: 34997252-34997252 on Build GRCh38
rs755598003 Chr.9: 34997273-34997273 on Build GRCh38
rs778677308 Chr.9: 34997165-34997165 on Build GRCh38
rs910724675 Chr.9: 34997331-34997331 on Build GRCh38
rs770118094 Chr.9: 34997332-34997332 on Build GRCh38
rs776856885 Chr.9: 34997229-34997229 on Build GRCh38
rs941097778 Chr.9: 34997247-34997247 on Build GRCh38
rs753416773 Chr.9: 34997281-34997281 on Build GRCh38
rs780509105 Chr.9: 34997339-34997339 on Build GRCh38
rs931820652 Chr.9: 34997337-34997337 on Build GRCh38
rs756893350 Chr.9: 34997286-34997286 on Build GRCh38
rs778591083 Chr.9: 34997294-34997294 on Build GRCh38
rs991590748 Chr.9: 34997205-34997205 on Build GRCh38
rs766874046 Chr.9: 34997262-34997262 on Build GRCh38
rs763519432 Chr.9: 34997278-34997278 on Build GRCh38

View Full Product Details