Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1614555-1614786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRAMP1
Entrez Gene ID
57585
Gene Name
cramped chromatin regulator homolog 1
Gene Aliases
CRAMP1L, HN1L, TCE4
UniGene
Hs.603676
Gene Chromosome Location
Chr.16: 1614640-1677908 on Build GRCh38
Transcripts
NM_020825.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6455093 c.106G>A p.A36T Chr.16: 1614745-1614745 on Build GRCh38
6455094 c.106G>A p.A36T Chr.16: 1614745-1614745 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs901281519 Chr.16: 1614576-1614576 on Build GRCh38
rs980268947 Chr.16: 1614678-1614678 on Build GRCh38
rs1005767706 Chr.16: 1614604-1614604 on Build GRCh38
rs1024453985 Chr.16: 1614663-1614663 on Build GRCh38
rs961528142 Chr.16: 1614626-1614626 on Build GRCh38
rs780609611 Chr.16: 1614603-1614603 on Build GRCh38
rs971145177 Chr.16: 1614673-1614673 on Build GRCh38
rs994373596 Chr.16: 1614629-1614629 on Build GRCh38
rs998695726 Chr.16: 1614581-1614581 on Build GRCh38
rs756460271 Chr.16: 1614596-1614596 on Build GRCh38
rs748220918 Chr.16: 1614738-1614738 on Build GRCh38
rs533763932 Chr.16: 1614575-1614575 on Build GRCh38
rs889799010 Chr.16: 1614591-1614591 on Build GRCh38
rs868231334 Chr.16: 1614745-1614745 on Build GRCh38
rs755456226 Chr.16: 1614691-1614691 on Build GRCh38
rs926923881 Chr.16: 1614695-1614695 on Build GRCh38
rs957076005 Chr.16: 1614748-1614748 on Build GRCh38
rs1028295962 Chr.16: 1614582-1614582 on Build GRCh38
rs778881967 Chr.16: 1614720-1614720 on Build GRCh38
rs754351402 Chr.16: 1614684-1614684 on Build GRCh38

View Full Product Details