Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176396374-176396645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL10
Entrez Gene ID
285598
Gene Name
ADP ribosylation factor like GTPase 10
Gene Aliases
ARL10A
UniGene
Hs.424552
Gene Chromosome Location
Chr.5: 176365468-176415085 on Build GRCh38
Transcripts
XM_011534529.2 XM_011534530.2 NM_001834.3 NM_007097.3 NR_045724.1 XM_017009026.1

Gene Symbol
CLTB
Entrez Gene ID
1212
Gene Name
clathrin light chain B
Gene Aliases
LCB
UniGene
Hs.484241
Gene Chromosome Location
Chr.5: 176392455-176416614 on Build GRCh38
Transcripts
XM_011534529.2 XM_011534530.2 NM_001834.3 NM_007097.3 NR_045724.1 XM_017009026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5578311 c.505A>T p.I169F Chr.5: 176396492-176396492 on Build GRCh38
6630772 c.501T>G p.A167A Chr.5: 176396496-176396496 on Build GRCh38
5928669 c.465-4C>T p.? Chr.5: 176396536-176396536 on Build GRCh38
1066020 c.510C>T p.I170I Chr.5: 176396487-176396487 on Build GRCh38
5413865 c.518+1G>T p.? Chr.5: 176396478-176396478 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751799129 Chr.5: 176396557-176396557 on Build GRCh38
rs377645084 Chr.5: 176396486-176396486 on Build GRCh38
rs748680853 Chr.5: 176396566-176396566 on Build GRCh38
rs753694498 Chr.5: 176396554-176396554 on Build GRCh38
rs550273130 Chr.5: 176396509-176396509 on Build GRCh38
rs143340859 Chr.5: 176396403-176396403 on Build GRCh38
rs527975677 Chr.5: 176396427-176396427 on Build GRCh38
rs371945626 Chr.5: 176396542-176396542 on Build GRCh38
rs374359977 Chr.5: 176396547-176396547 on Build GRCh38
rs991439684 Chr.5: 176396536-176396536 on Build GRCh38
rs781485327 Chr.5: 176396447-176396448 on Build GRCh38
rs746060352 Chr.5: 176396439-176396439 on Build GRCh38
rs374264390 Chr.5: 176396622-176396623 on Build GRCh38
rs373717718 Chr.5: 176396475-176396475 on Build GRCh38
rs770692413 Chr.5: 176396416-176396416 on Build GRCh38
rs774149649 Chr.5: 176396500-176396500 on Build GRCh38
rs369257313 Chr.5: 176396468-176396468 on Build GRCh38
rs780185838 Chr.5: 176396556-176396556 on Build GRCh38
rs776517011 Chr.5: 176396487-176396487 on Build GRCh38
rs1040731033 Chr.5: 176396520-176396520 on Build GRCh38
rs757062644 Chr.5: 176396555-176396555 on Build GRCh38
rs535983644 Chr.5: 176396624-176396624 on Build GRCh38
rs564656616 Chr.5: 176396443-176396443 on Build GRCh38
rs752659726 Chr.5: 176396528-176396528 on Build GRCh38
rs957943442 Chr.5: 176396578-176396578 on Build GRCh38
rs376796902 Chr.5: 176396607-176396608 on Build GRCh38
rs748649397 Chr.5: 176396477-176396477 on Build GRCh38
rs981755988 Chr.5: 176396606-176396607 on Build GRCh38
rs755060024 Chr.5: 176396565-176396565 on Build GRCh38
rs778437501 Chr.5: 176396580-176396580 on Build GRCh38
rs776492275 Chr.5: 176396581-176396581 on Build GRCh38
rs745926137 Chr.5: 176396550-176396550 on Build GRCh38
rs770649500 Chr.5: 176396494-176396494 on Build GRCh38
rs375585817 Chr.5: 176396565-176396565 on Build GRCh38
rs770261797 Chr.5: 176396574-176396574 on Build GRCh38
rs1010750097 Chr.5: 176396607-176396607 on Build GRCh38
rs770160355 Chr.5: 176396480-176396480 on Build GRCh38
rs889546779 Chr.5: 176396504-176396504 on Build GRCh38
rs552862988 Chr.5: 176396428-176396428 on Build GRCh38
rs940392946 Chr.5: 176396623-176396623 on Build GRCh38
rs760569600 Chr.5: 176396529-176396529 on Build GRCh38

View Full Product Details