Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 134861866-134862123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACMSD
Entrez Gene ID
130013
Gene Name
aminocarboxymuconate semialdehyde decarboxylase
Gene Aliases
-
UniGene
Hs.655728
Gene Chromosome Location
Chr.2: 134838319-134902906 on Build GRCh38
Transcripts
NM_001307983.1 NM_138326.2 XM_005263586.4 XM_005263588.4 XM_005263589.4 XM_005263590.4 XM_011510592.2 XM_017003325.1 XM_017003326.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1717944 c.230C>T p.P77L Chr.2: 134861999-134861999 on Build GRCh38
6257651 c.200-1G>T p.? Chr.2: 134861968-134861968 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755769626 Chr.2: 134862054-134862054 on Build GRCh38
rs1008784833 Chr.2: 134861986-134861986 on Build GRCh38
rs757703384 Chr.2: 134862062-134862062 on Build GRCh38
rs917542427 Chr.2: 134862097-134862097 on Build GRCh38
rs373563280 Chr.2: 134862061-134862061 on Build GRCh38
rs775065929 Chr.2: 134861947-134861947 on Build GRCh38
rs941480228 Chr.2: 134861894-134861894 on Build GRCh38
rs751500402 Chr.2: 134862037-134862037 on Build GRCh38
rs566947149 Chr.2: 134861892-134861892 on Build GRCh38
rs556103429 Chr.2: 134862075-134862075 on Build GRCh38
rs752190855 Chr.2: 134862044-134862044 on Build GRCh38
rs138372803 Chr.2: 134862043-134862043 on Build GRCh38
rs766298298 Chr.2: 134862027-134862027 on Build GRCh38
rs1038435491 Chr.2: 134861897-134861897 on Build GRCh38
rs190193236 Chr.2: 134861909-134861909 on Build GRCh38
rs759624053 Chr.2: 134861957-134861957 on Build GRCh38
rs749723124 Chr.2: 134861933-134861933 on Build GRCh38
rs771552596 Chr.2: 134861935-134861935 on Build GRCh38
rs150911787 Chr.2: 134861930-134861930 on Build GRCh38
rs760985297 Chr.2: 134862009-134862009 on Build GRCh38
rs772208512 Chr.2: 134861962-134861962 on Build GRCh38
rs777351762 Chr.2: 134862057-134862057 on Build GRCh38
rs754385456 Chr.2: 134862038-134862038 on Build GRCh38
rs75020600 Chr.2: 134861915-134861915 on Build GRCh38
rs750192483 Chr.2: 134862015-134862015 on Build GRCh38
rs762793455 Chr.2: 134862018-134862018 on Build GRCh38
rs775523958 Chr.2: 134861997-134861997 on Build GRCh38
rs989660898 Chr.2: 134862091-134862091 on Build GRCh38

View Full Product Details