Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 98209587-98209844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
R3HCC1L
Entrez Gene ID
27291
Gene Name
R3H domain and coiled-coil containing 1 like
Gene Aliases
C10orf28, GIDRP86, GIDRP88, PSORT
UniGene
Hs.419800
Gene Chromosome Location
Chr.10: 98134608-98244897 on Build GRCh38
Transcripts
NM_001256619.1 NM_001256620.1 NM_001256621.1 NM_014472.4 NM_138469.2 XM_011539639.1 XM_011539640.1 XM_011539641.1 XM_011539642.1 XM_011539643.1 XM_011539644.1 XM_011539645.2 XM_011539646.1 XM_011539647.1 XM_011539648.1 XM_011539649.1 XM_017016074.1 XM_017016075.1 XM_017016076.1 XM_017016077.1 XM_017016078.1 XM_017016079.1 XM_017016080.1 XM_017016081.1 XM_017016082.1 XM_017016083.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3935265 c.1609G>T p.G537W Chr.10: 98209723-98209723 on Build GRCh38
399621 c.1528G>T p.G510C Chr.10: 98209642-98209642 on Build GRCh38
6761074 c.1660G>A p.E554K Chr.10: 98209774-98209774 on Build GRCh38
922099 c.1672G>A p.E558K Chr.10: 98209786-98209786 on Build GRCh38
4998354 c.1558C>A p.H520N Chr.10: 98209672-98209672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138288770 Chr.10: 98209733-98209733 on Build GRCh38
rs765186392 Chr.10: 98209767-98209767 on Build GRCh38
rs772821159 Chr.10: 98209741-98209742 on Build GRCh38
rs775543308 Chr.10: 98209663-98209663 on Build GRCh38
rs531584288 Chr.10: 98209653-98209653 on Build GRCh38
rs147959217 Chr.10: 98209661-98209661 on Build GRCh38
rs1042896101 Chr.10: 98209625-98209625 on Build GRCh38
rs961107798 Chr.10: 98209780-98209780 on Build GRCh38
rs768744727 Chr.10: 98209754-98209754 on Build GRCh38
rs552450363 Chr.10: 98209642-98209642 on Build GRCh38
rs760243702 Chr.10: 98209761-98209763 on Build GRCh38
rs761872252 Chr.10: 98209717-98209718 on Build GRCh38
rs775557593 Chr.10: 98209785-98209785 on Build GRCh38
rs370785776 Chr.10: 98209817-98209817 on Build GRCh38
rs771903405 Chr.10: 98209722-98209723 on Build GRCh38
rs771284727 Chr.10: 98209656-98209656 on Build GRCh38
rs765053075 Chr.10: 98209689-98209689 on Build GRCh38
rs74153524 Chr.10: 98209633-98209633 on Build GRCh38
rs754054422 Chr.10: 98209759-98209759 on Build GRCh38
rs77532805 Chr.10: 98209682-98209682 on Build GRCh38
rs34494334 Chr.10: 98209718-98209718 on Build GRCh38
rs563014187 Chr.10: 98209698-98209698 on Build GRCh38
rs749770683 Chr.10: 98209722-98209722 on Build GRCh38
rs762776274 Chr.10: 98209810-98209810 on Build GRCh38
rs200555053 Chr.10: 98209816-98209816 on Build GRCh38
rs145569032 Chr.10: 98209731-98209731 on Build GRCh38
rs760742984 Chr.10: 98209674-98209674 on Build GRCh38
rs142377469 Chr.10: 98209632-98209632 on Build GRCh38
rs771166083 Chr.10: 98209699-98209699 on Build GRCh38
rs778283837 Chr.10: 98209650-98209650 on Build GRCh38
rs199945129 Chr.10: 98209658-98209658 on Build GRCh38
rs369504502 Chr.10: 98209627-98209627 on Build GRCh38
rs1056868156 Chr.10: 98209727-98209727 on Build GRCh38
rs750156034 Chr.10: 98209690-98209690 on Build GRCh38
rs35122894 Chr.10: 98209750-98209750 on Build GRCh38
rs140585440 Chr.10: 98209680-98209680 on Build GRCh38
rs757777932 Chr.10: 98209818-98209818 on Build GRCh38
rs763956118 Chr.10: 98209677-98209677 on Build GRCh38
rs574187705 Chr.10: 98209646-98209646 on Build GRCh38
rs11189513 Chr.10: 98209811-98209811 on Build GRCh38
rs779695870 Chr.10: 98209639-98209639 on Build GRCh38
rs776475971 Chr.10: 98209761-98209761 on Build GRCh38
rs753583463 Chr.10: 98209621-98209621 on Build GRCh38
rs371232521 Chr.10: 98209712-98209712 on Build GRCh38
rs76898723 Chr.10: 98209678-98209678 on Build GRCh38
rs1027086469 Chr.10: 98209726-98209726 on Build GRCh38
rs766153187 Chr.10: 98209792-98209816 on Build GRCh38
rs747954459 Chr.10: 98209660-98209660 on Build GRCh38
rs976380480 Chr.10: 98209631-98209631 on Build GRCh38
rs751181899 Chr.10: 98209812-98209812 on Build GRCh38
rs758028142 Chr.10: 98209638-98209638 on Build GRCh38
rs747085556 Chr.10: 98209753-98209753 on Build GRCh38
rs761792896 Chr.10: 98209764-98209764 on Build GRCh38
rs772287059 Chr.10: 98209745-98209745 on Build GRCh38
rs148867373 Chr.10: 98209741-98209741 on Build GRCh38
rs754487235 Chr.10: 98209813-98209813 on Build GRCh38

View Full Product Details